Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 18851424-18851970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD20A9P
Entrez Gene ID
284232
Gene Name
ankyrin repeat domain 20 family member A9, pseudogene
Gene Aliases
-
UniGene
Hs.679496
Gene Chromosome Location
Chr.13: 18834403-18871969 on Build GRCh38
Transcripts
NR_027995.1

SNP Detail

SNP ID Chromosome Location
rs902192706 Chr.13: 18851525-18851525 on Build GRCh38
rs538448200 Chr.13: 18851598-18851598 on Build GRCh38
rs34750429 Chr.13: 18851944-18851945 on Build GRCh38
rs1013035057 Chr.13: 18851582-18851582 on Build GRCh38
rs978363650 Chr.13: 18851924-18851924 on Build GRCh38
rs952878824 Chr.13: 18851612-18851612 on Build GRCh38
rs547715918 Chr.13: 18851874-18851874 on Build GRCh38
rs1049554095 Chr.13: 18851663-18851663 on Build GRCh38
rs35239687 Chr.13: 18851729-18851729 on Build GRCh38
rs145227140 Chr.13: 18851669-18851669 on Build GRCh38
rs565224488 Chr.13: 18851711-18851711 on Build GRCh38
rs961802282 Chr.13: 18851780-18851780 on Build GRCh38
rs115238013 Chr.13: 18851658-18851658 on Build GRCh38
rs541227550 Chr.13: 18851799-18851799 on Build GRCh38
rs12859891 Chr.13: 18851558-18851558 on Build GRCh38
rs1006162957 Chr.13: 18851760-18851760 on Build GRCh38
rs532531673 Chr.13: 18851868-18851868 on Build GRCh38
rs1029679107 Chr.13: 18851809-18851809 on Build GRCh38
rs548575725 Chr.13: 18851460-18851460 on Build GRCh38
rs939023036 Chr.13: 18851716-18851716 on Build GRCh38
rs1042415537 Chr.13: 18851479-18851480 on Build GRCh38
rs566891936 Chr.13: 18851502-18851502 on Build GRCh38
rs946432346 Chr.13: 18851696-18851696 on Build GRCh38
rs946555756 Chr.13: 18851900-18851900 on Build GRCh38
rs1056095340 Chr.13: 18851727-18851727 on Build GRCh38
rs537970456 Chr.13: 18851545-18851545 on Build GRCh38
rs960314707 Chr.13: 18851570-18851570 on Build GRCh38
rs576335948 Chr.13: 18851684-18851684 on Build GRCh38
rs751151592 Chr.13: 18851736-18851736 on Build GRCh38
rs9510200 Chr.13: 18851745-18851745 on Build GRCh38
rs566010585 Chr.13: 18851882-18851882 on Build GRCh38
rs150020638 Chr.13: 18851564-18851564 on Build GRCh38
rs373369581 Chr.13: 18851675-18851675 on Build GRCh38
rs902289500 Chr.13: 18851710-18851710 on Build GRCh38
rs754635373 Chr.13: 18851818-18851818 on Build GRCh38
rs191809073 Chr.13: 18851621-18851621 on Build GRCh38
rs893522680 Chr.13: 18851446-18851446 on Build GRCh38
rs1010661913 Chr.13: 18851458-18851458 on Build GRCh38
rs1015673721 Chr.13: 18851778-18851783 on Build GRCh38
rs896031801 Chr.13: 18851731-18851731 on Build GRCh38
rs908644259 Chr.13: 18851635-18851635 on Build GRCh38
rs77069454 Chr.13: 18851657-18851657 on Build GRCh38
rs796257859 Chr.13: 18851753-18851753 on Build GRCh38
rs1023066246 Chr.13: 18851747-18851747 on Build GRCh38
rs559477911 Chr.13: 18851850-18851850 on Build GRCh38
rs1023090815 Chr.13: 18851599-18851599 on Build GRCh38
rs915164203 Chr.13: 18851881-18851881 on Build GRCh38
rs34103693 Chr.13: 18851605-18851605 on Build GRCh38
rs112337315 Chr.13: 18851791-18851791 on Build GRCh38
rs976712061 Chr.13: 18851781-18851781 on Build GRCh38
rs183011292 Chr.13: 18851702-18851702 on Build GRCh38

View Full Product Details