Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72679861-72680409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIGD2B
Entrez Gene ID
123346
Gene Name
HIG1 hypoxia inducible domain family member 2B
Gene Aliases
HIGD2BP
UniGene
Hs.434111
Gene Chromosome Location
Chr.15: 72675782-72686149 on Build GRCh38
Transcripts
NR_002780.1

SNP Detail

SNP ID Chromosome Location
rs961902115 Chr.15: 72680146-72680146 on Build GRCh38
rs911693523 Chr.15: 72680004-72680004 on Build GRCh38
rs957465588 Chr.15: 72680356-72680356 on Build GRCh38
rs975927270 Chr.15: 72680374-72680374 on Build GRCh38
rs571231055 Chr.15: 72680100-72680100 on Build GRCh38
rs911977962 Chr.15: 72680016-72680016 on Build GRCh38
rs188972311 Chr.15: 72680147-72680147 on Build GRCh38
rs367962433 Chr.15: 72680012-72680012 on Build GRCh38
rs920529682 Chr.15: 72680185-72680185 on Build GRCh38
rs114436764 Chr.15: 72680143-72680143 on Build GRCh38
rs1036090119 Chr.15: 72680022-72680022 on Build GRCh38
rs960063699 Chr.15: 72680082-72680082 on Build GRCh38
rs907748141 Chr.15: 72679964-72679964 on Build GRCh38
rs973299049 Chr.15: 72680178-72680178 on Build GRCh38
rs1014499022 Chr.15: 72680223-72680223 on Build GRCh38
rs1034070942 Chr.15: 72680072-72680072 on Build GRCh38
rs75531053 Chr.15: 72679956-72679956 on Build GRCh38
rs967550590 Chr.15: 72679893-72679893 on Build GRCh38
rs745326241 Chr.15: 72679969-72679969 on Build GRCh38
rs898927872 Chr.15: 72680023-72680023 on Build GRCh38
rs16957238 Chr.15: 72680206-72680206 on Build GRCh38
rs760503044 Chr.15: 72679916-72679916 on Build GRCh38
rs1014045635 Chr.15: 72680083-72680083 on Build GRCh38
rs973354123 Chr.15: 72679977-72679977 on Build GRCh38
rs1033029461 Chr.15: 72680314-72680314 on Build GRCh38
rs1052155019 Chr.15: 72680002-72680002 on Build GRCh38
rs892668976 Chr.15: 72680035-72680035 on Build GRCh38
rs780099983 Chr.15: 72680348-72680349 on Build GRCh38
rs961954231 Chr.15: 72679965-72679965 on Build GRCh38
rs959869848 Chr.15: 72679913-72679913 on Build GRCh38
rs569411347 Chr.15: 72680051-72680051 on Build GRCh38
rs778035590 Chr.15: 72679994-72679994 on Build GRCh38
rs926044606 Chr.15: 72679947-72679947 on Build GRCh38
rs1011099667 Chr.15: 72680037-72680037 on Build GRCh38
rs921293894 Chr.15: 72679987-72679987 on Build GRCh38
rs998999047 Chr.15: 72680292-72680292 on Build GRCh38
rs183641653 Chr.15: 72680025-72680025 on Build GRCh38
rs1024522967 Chr.15: 72680276-72680276 on Build GRCh38
rs773857358 Chr.15: 72679950-72679950 on Build GRCh38
rs775259497 Chr.15: 72680049-72680049 on Build GRCh38
rs538253000 Chr.15: 72680283-72680283 on Build GRCh38
rs1001618637 Chr.15: 72680055-72680055 on Build GRCh38
rs566025883 Chr.15: 72680166-72680166 on Build GRCh38
rs549503595 Chr.15: 72680048-72680048 on Build GRCh38

View Full Product Details