Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75730115-75730686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1P35
Entrez Gene ID
100128285
Gene Name
dynamin 1 pseudogene 35
Gene Aliases
DNM1DN8-2, DNM1DN8.2, DNM1DN8@, FKSG88
UniGene
Hs.604066
Gene Chromosome Location
Chr.15: 75727670-75740077 on Build GRCh38
Transcripts
NR_024595.3

SNP Detail

SNP ID Chromosome Location
rs568855261 Chr.15: 75730572-75730572 on Build GRCh38
rs376817697 Chr.15: 75730339-75730339 on Build GRCh38
rs535167049 Chr.15: 75730643-75730643 on Build GRCh38
rs962902103 Chr.15: 75730144-75730144 on Build GRCh38
rs72732879 Chr.15: 75730343-75730343 on Build GRCh38
rs796556113 Chr.15: 75730338-75730338 on Build GRCh38
rs1044334002 Chr.15: 75730341-75730342 on Build GRCh38
rs574713904 Chr.15: 75730307-75730307 on Build GRCh38
rs569994244 Chr.15: 75730478-75730478 on Build GRCh38
rs72498385 Chr.15: 75730340-75730341 on Build GRCh38
rs139257603 Chr.15: 75730394-75730394 on Build GRCh38
rs1028488712 Chr.15: 75730163-75730163 on Build GRCh38
rs987319801 Chr.15: 75730204-75730204 on Build GRCh38
rs898834320 Chr.15: 75730343-75730344 on Build GRCh38
rs531039607 Chr.15: 75730342-75730342 on Build GRCh38
rs891965924 Chr.15: 75730613-75730613 on Build GRCh38
rs765508919 Chr.15: 75730581-75730581 on Build GRCh38
rs954232131 Chr.15: 75730163-75730164 on Build GRCh38
rs968698499 Chr.15: 75730417-75730417 on Build GRCh38
rs566042899 Chr.15: 75730614-75730614 on Build GRCh38
rs764564324 Chr.15: 75730468-75730468 on Build GRCh38
rs912631019 Chr.15: 75730293-75730293 on Build GRCh38
rs193167370 Chr.15: 75730601-75730601 on Build GRCh38
rs995637629 Chr.15: 75730162-75730165 on Build GRCh38
rs1015814309 Chr.15: 75730137-75730137 on Build GRCh38
rs150811395 Chr.15: 75730166-75730166 on Build GRCh38
rs1008372631 Chr.15: 75730351-75730351 on Build GRCh38
rs201138544 Chr.15: 75730338-75730339 on Build GRCh38
rs528839698 Chr.15: 75730561-75730561 on Build GRCh38
rs1020131434 Chr.15: 75730370-75730370 on Build GRCh38
rs28478620 Chr.15: 75730582-75730582 on Build GRCh38
rs191020937 Chr.15: 75730241-75730241 on Build GRCh38
rs35510810 Chr.15: 75730339-75730339 on Build GRCh38
rs369229657 Chr.15: 75730338-75730338 on Build GRCh38
rs182843791 Chr.15: 75730627-75730627 on Build GRCh38
rs571837318 Chr.15: 75730339-75730340 on Build GRCh38
rs147280585 Chr.15: 75730555-75730555 on Build GRCh38
rs28727341 Chr.15: 75730420-75730420 on Build GRCh38
rs563592392 Chr.15: 75730341-75730341 on Build GRCh38
rs968727883 Chr.15: 75730644-75730644 on Build GRCh38
rs28587788 Chr.15: 75730458-75730458 on Build GRCh38
rs956254962 Chr.15: 75730323-75730323 on Build GRCh38
rs992644403 Chr.15: 75730462-75730462 on Build GRCh38
rs931632053 Chr.15: 75730573-75730573 on Build GRCh38
rs938530435 Chr.15: 75730451-75730451 on Build GRCh38
rs550550544 Chr.15: 75730571-75730571 on Build GRCh38
rs890016703 Chr.15: 75730584-75730584 on Build GRCh38
rs201828954 Chr.15: 75730604-75730604 on Build GRCh38
rs564382530 Chr.15: 75730346-75730346 on Build GRCh38
rs748848376 Chr.15: 75730225-75730225 on Build GRCh38
rs1028564109 Chr.15: 75730345-75730345 on Build GRCh38
rs995751776 Chr.15: 75730344-75730344 on Build GRCh38
rs1021674308 Chr.15: 75730397-75730397 on Build GRCh38
rs533999046 Chr.15: 75730652-75730652 on Build GRCh38
rs550096462 Chr.15: 75730421-75730421 on Build GRCh38
rs956114122 Chr.15: 75730376-75730376 on Build GRCh38

View Full Product Details