Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75731857-75732398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1P35
Entrez Gene ID
100128285
Gene Name
dynamin 1 pseudogene 35
Gene Aliases
DNM1DN8-2, DNM1DN8.2, DNM1DN8@, FKSG88
UniGene
Hs.604066
Gene Chromosome Location
Chr.15: 75727670-75740077 on Build GRCh38
Transcripts
NR_024595.3

SNP Detail

SNP ID Chromosome Location
rs56851614 Chr.15: 75731906-75731907 on Build GRCh38
rs1040282503 Chr.15: 75732052-75732052 on Build GRCh38
rs797017508 Chr.15: 75731882-75731883 on Build GRCh38
rs903686711 Chr.15: 75732321-75732321 on Build GRCh38
rs574149202 Chr.15: 75732178-75732178 on Build GRCh38
rs991754562 Chr.15: 75731958-75731958 on Build GRCh38
rs189854152 Chr.15: 75732028-75732028 on Build GRCh38
rs1040714754 Chr.15: 75732233-75732233 on Build GRCh38
rs943366500 Chr.15: 75732217-75732217 on Build GRCh38
rs376602178 Chr.15: 75732331-75732331 on Build GRCh38
rs1000537020 Chr.15: 75732109-75732109 on Build GRCh38
rs1042206737 Chr.15: 75732289-75732289 on Build GRCh38
rs139874054 Chr.15: 75731893-75731896 on Build GRCh38
rs34544213 Chr.15: 75731881-75731882 on Build GRCh38
rs796784054 Chr.15: 75731906-75731907 on Build GRCh38
rs530259186 Chr.15: 75732322-75732322 on Build GRCh38
rs397854253 Chr.15: 75731905-75731906 on Build GRCh38
rs796252110 Chr.15: 75731882-75731885 on Build GRCh38
rs952089359 Chr.15: 75732183-75732192 on Build GRCh38
rs930657965 Chr.15: 75732005-75732005 on Build GRCh38
rs397836574 Chr.15: 75731883-75731884 on Build GRCh38
rs572988848 Chr.15: 75732033-75732033 on Build GRCh38
rs773163327 Chr.15: 75731885-75731886 on Build GRCh38
rs766422733 Chr.15: 75731884-75731885 on Build GRCh38
rs556855165 Chr.15: 75732223-75732223 on Build GRCh38
rs1033275047 Chr.15: 75731901-75731901 on Build GRCh38
rs972145933 Chr.15: 75731974-75731974 on Build GRCh38
rs557774045 Chr.15: 75731989-75731989 on Build GRCh38
rs544494284 Chr.15: 75732186-75732186 on Build GRCh38
rs903573864 Chr.15: 75732068-75732068 on Build GRCh38
rs28695716 Chr.15: 75732356-75732356 on Build GRCh38
rs777063725 Chr.15: 75732050-75732050 on Build GRCh38
rs919319533 Chr.15: 75731998-75731998 on Build GRCh38
rs76926910 Chr.15: 75731951-75731951 on Build GRCh38
rs1009381765 Chr.15: 75732053-75732053 on Build GRCh38
rs917441091 Chr.15: 75731969-75731969 on Build GRCh38
rs578250940 Chr.15: 75732237-75732237 on Build GRCh38
rs923157151 Chr.15: 75732236-75732236 on Build GRCh38
rs1049496322 Chr.15: 75732006-75732006 on Build GRCh38
rs1013615771 Chr.15: 75732143-75732143 on Build GRCh38
rs959063015 Chr.15: 75732112-75732112 on Build GRCh38
rs910470147 Chr.15: 75732017-75732017 on Build GRCh38
rs1042176426 Chr.15: 75732064-75732064 on Build GRCh38
rs550023410 Chr.15: 75731921-75731921 on Build GRCh38
rs555658156 Chr.15: 75732173-75732173 on Build GRCh38
rs762293744 Chr.15: 75732051-75732051 on Build GRCh38
rs545020079 Chr.15: 75732283-75732283 on Build GRCh38
rs371863613 Chr.15: 75732285-75732285 on Build GRCh38
rs561612644 Chr.15: 75732147-75732147 on Build GRCh38
rs59136529 Chr.15: 75731903-75731906 on Build GRCh38
rs539409661 Chr.15: 75731952-75731952 on Build GRCh38
rs766571710 Chr.15: 75732323-75732323 on Build GRCh38
rs910575322 Chr.15: 75732205-75732205 on Build GRCh38
rs368065036 Chr.15: 75732111-75732111 on Build GRCh38

View Full Product Details