Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75738274-75738828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1P35
Entrez Gene ID
100128285
Gene Name
dynamin 1 pseudogene 35
Gene Aliases
DNM1DN8-2, DNM1DN8.2, DNM1DN8@, FKSG88
UniGene
Hs.604066
Gene Chromosome Location
Chr.15: 75727670-75740077 on Build GRCh38
Transcripts
NR_024595.3

SNP Detail

SNP ID Chromosome Location
rs1026319823 Chr.15: 75738476-75738476 on Build GRCh38
rs1006664417 Chr.15: 75738643-75738643 on Build GRCh38
rs928787300 Chr.15: 75738560-75738560 on Build GRCh38
rs530958768 Chr.15: 75738609-75738609 on Build GRCh38
rs562353842 Chr.15: 75738593-75738593 on Build GRCh38
rs929575840 Chr.15: 75738773-75738773 on Build GRCh38
rs534711356 Chr.15: 75738629-75738629 on Build GRCh38
rs527434541 Chr.15: 75738710-75738710 on Build GRCh38
rs568943123 Chr.15: 75738375-75738375 on Build GRCh38
rs1047816793 Chr.15: 75738394-75738394 on Build GRCh38
rs773314420 Chr.15: 75738307-75738307 on Build GRCh38
rs984783647 Chr.15: 75738508-75738508 on Build GRCh38
rs370413743 Chr.15: 75738606-75738606 on Build GRCh38
rs556912180 Chr.15: 75738622-75738622 on Build GRCh38
rs766249716 Chr.15: 75738502-75738503 on Build GRCh38
rs970757176 Chr.15: 75738676-75738676 on Build GRCh38
rs946478744 Chr.15: 75738753-75738753 on Build GRCh38
rs956206520 Chr.15: 75738719-75738719 on Build GRCh38
rs771105396 Chr.15: 75738395-75738395 on Build GRCh38
rs981720272 Chr.15: 75738703-75738703 on Build GRCh38
rs924993087 Chr.15: 75738328-75738328 on Build GRCh38
rs929436627 Chr.15: 75738393-75738393 on Build GRCh38
rs747074778 Chr.15: 75738384-75738384 on Build GRCh38
rs917694162 Chr.15: 75738637-75738653 on Build GRCh38
rs942992275 Chr.15: 75738776-75738776 on Build GRCh38
rs900484680 Chr.15: 75738424-75738424 on Build GRCh38
rs952058845 Chr.15: 75738484-75738484 on Build GRCh38
rs376919305 Chr.15: 75738792-75738792 on Build GRCh38
rs970323704 Chr.15: 75738550-75738550 on Build GRCh38
rs552255594 Chr.15: 75738323-75738323 on Build GRCh38
rs887822207 Chr.15: 75738398-75738398 on Build GRCh38
rs901689614 Chr.15: 75738625-75738625 on Build GRCh38
rs887845944 Chr.15: 75738635-75738635 on Build GRCh38
rs990519370 Chr.15: 75738532-75738532 on Build GRCh38
rs913681757 Chr.15: 75738742-75738742 on Build GRCh38
rs901974371 Chr.15: 75738431-75738431 on Build GRCh38
rs147020979 Chr.15: 75738305-75738305 on Build GRCh38
rs759402648 Chr.15: 75738662-75738662 on Build GRCh38
rs987896764 Chr.15: 75738586-75738586 on Build GRCh38
rs753412665 Chr.15: 75738658-75738658 on Build GRCh38
rs533230537 Chr.15: 75738785-75738785 on Build GRCh38
rs1030632671 Chr.15: 75738452-75738452 on Build GRCh38
rs1048396501 Chr.15: 75738612-75738612 on Build GRCh38
rs918255930 Chr.15: 75738756-75738756 on Build GRCh38
rs769724027 Chr.15: 75738377-75738377 on Build GRCh38
rs748182658 Chr.15: 75738320-75738320 on Build GRCh38
rs12904654 Chr.15: 75738652-75738652 on Build GRCh38
rs927730152 Chr.15: 75738308-75738308 on Build GRCh38
rs758590254 Chr.15: 75738360-75738360 on Build GRCh38
rs768145060 Chr.15: 75738729-75738729 on Build GRCh38
rs558188212 Chr.15: 75738454-75738454 on Build GRCh38
rs942146353 Chr.15: 75738803-75738803 on Build GRCh38
rs997451635 Chr.15: 75738444-75738444 on Build GRCh38
rs947587374 Chr.15: 75738673-75738673 on Build GRCh38
rs141399305 Chr.15: 75738304-75738304 on Build GRCh38
rs999443976 Chr.15: 75738696-75738696 on Build GRCh38
rs542803460 Chr.15: 75738739-75738739 on Build GRCh38
rs773073873 Chr.15: 75738472-75738472 on Build GRCh38
rs916242883 Chr.15: 75738347-75738347 on Build GRCh38
rs957819309 Chr.15: 75738521-75738521 on Build GRCh38
rs949003844 Chr.15: 75738363-75738363 on Build GRCh38
rs185446057 Chr.15: 75738423-75738423 on Build GRCh38
rs1043384166 Chr.15: 75738607-75738607 on Build GRCh38
rs868120112 Chr.15: 75738768-75738768 on Build GRCh38
rs7181595 Chr.15: 75738667-75738667 on Build GRCh38
rs1054756471 Chr.15: 75738345-75738345 on Build GRCh38
rs901743448 Chr.15: 75738787-75738787 on Build GRCh38
rs957849558 Chr.15: 75738713-75738713 on Build GRCh38
rs892139731 Chr.15: 75738338-75738338 on Build GRCh38
rs747900540 Chr.15: 75738337-75738337 on Build GRCh38
rs982054375 Chr.15: 75738552-75738552 on Build GRCh38
rs753753532 Chr.15: 75738319-75738319 on Build GRCh38
rs566863516 Chr.15: 75738568-75738568 on Build GRCh38
rs560774839 Chr.15: 75738755-75738755 on Build GRCh38
rs1007832280 Chr.15: 75738408-75738408 on Build GRCh38
rs570582779 Chr.15: 75738600-75738600 on Build GRCh38
rs868021329 Chr.15: 75738637-75738637 on Build GRCh38
rs943297572 Chr.15: 75738616-75738616 on Build GRCh38
rs542200423 Chr.15: 75738372-75738372 on Build GRCh38
rs1023664149 Chr.15: 75738537-75738537 on Build GRCh38
rs1026373948 Chr.15: 75738632-75738632 on Build GRCh38
rs772707682 Chr.15: 75738627-75738627 on Build GRCh38

View Full Product Details