Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75739781-75740342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1P35
Entrez Gene ID
100128285
Gene Name
dynamin 1 pseudogene 35
Gene Aliases
DNM1DN8-2, DNM1DN8.2, DNM1DN8@, FKSG88
UniGene
Hs.604066
Gene Chromosome Location
Chr.15: 75727670-75740077 on Build GRCh38
Transcripts
NR_024595.3

SNP Detail

SNP ID Chromosome Location
rs1029662188 Chr.15: 75739868-75739868 on Build GRCh38
rs377753791 Chr.15: 75739944-75739944 on Build GRCh38
rs867628667 Chr.15: 75739897-75739897 on Build GRCh38
rs979263191 Chr.15: 75740062-75740062 on Build GRCh38
rs1001182285 Chr.15: 75740016-75740016 on Build GRCh38
rs28688252 Chr.15: 75739898-75739898 on Build GRCh38
rs752986947 Chr.15: 75739815-75739815 on Build GRCh38
rs922887718 Chr.15: 75740119-75740119 on Build GRCh38
rs536786624 Chr.15: 75739908-75739908 on Build GRCh38
rs554194172 Chr.15: 75739899-75739899 on Build GRCh38
rs926031252 Chr.15: 75740063-75740063 on Build GRCh38
rs1020125507 Chr.15: 75740050-75740050 on Build GRCh38
rs138496203 Chr.15: 75739884-75739885 on Build GRCh38
rs942683010 Chr.15: 75739896-75739896 on Build GRCh38
rs746705711 Chr.15: 75740255-75740255 on Build GRCh38
rs898894349 Chr.15: 75740260-75740260 on Build GRCh38
rs184221310 Chr.15: 75739904-75739904 on Build GRCh38
rs946104815 Chr.15: 75739878-75739878 on Build GRCh38
rs964409757 Chr.15: 75740076-75740076 on Build GRCh38
rs886224593 Chr.15: 75739962-75739962 on Build GRCh38
rs908395951 Chr.15: 75740102-75740102 on Build GRCh38
rs996481668 Chr.15: 75739834-75739834 on Build GRCh38
rs1009176496 Chr.15: 75740289-75740289 on Build GRCh38
rs28541422 Chr.15: 75740302-75740302 on Build GRCh38
rs188174424 Chr.15: 75740130-75740130 on Build GRCh38
rs34557014 Chr.15: 75740298-75740299 on Build GRCh38
rs978825159 Chr.15: 75739880-75739880 on Build GRCh38
rs908722309 Chr.15: 75740190-75740190 on Build GRCh38
rs765690019 Chr.15: 75739907-75739907 on Build GRCh38
rs200760317 Chr.15: 75740120-75740121 on Build GRCh38
rs932388304 Chr.15: 75740220-75740220 on Build GRCh38
rs138189399 Chr.15: 75740249-75740249 on Build GRCh38
rs568761001 Chr.15: 75740308-75740308 on Build GRCh38
rs34605750 Chr.15: 75739942-75739942 on Build GRCh38
rs552932979 Chr.15: 75740121-75740121 on Build GRCh38
rs118125648 Chr.15: 75739895-75739895 on Build GRCh38
rs868250990 Chr.15: 75740140-75740140 on Build GRCh38
rs28651447 Chr.15: 75739804-75739804 on Build GRCh38
rs879396520 Chr.15: 75739902-75739902 on Build GRCh38
rs898837009 Chr.15: 75740007-75740007 on Build GRCh38
rs531822921 Chr.15: 75740189-75740189 on Build GRCh38
rs1014129906 Chr.15: 75740047-75740047 on Build GRCh38
rs899515059 Chr.15: 75739819-75739819 on Build GRCh38
rs890773381 Chr.15: 75740262-75740262 on Build GRCh38
rs574716949 Chr.15: 75740101-75740101 on Build GRCh38
rs959932676 Chr.15: 75740040-75740040 on Build GRCh38
rs913346840 Chr.15: 75739849-75739849 on Build GRCh38
rs1034290834 Chr.15: 75740019-75740019 on Build GRCh38
rs941507891 Chr.15: 75740228-75740228 on Build GRCh38
rs552952095 Chr.15: 75740051-75740051 on Build GRCh38
rs1005040762 Chr.15: 75739980-75739980 on Build GRCh38
rs541224579 Chr.15: 75739992-75739992 on Build GRCh38
rs1037419656 Chr.15: 75739998-75739998 on Build GRCh38
rs1022301327 Chr.15: 75739900-75739900 on Build GRCh38
rs920977664 Chr.15: 75740132-75740132 on Build GRCh38
rs888183908 Chr.15: 75739827-75739827 on Build GRCh38

View Full Product Details