Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89971584-89972159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPBD1
Entrez Gene ID
92806
Gene Name
CENPB DNA-binding domain containing 1
Gene Aliases
-
UniGene
Hs.513832
Gene Chromosome Location
Chr.16: 89969775-89972832 on Build GRCh38
Transcripts
NM_145039.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063706 c.227C>T p.A76V Chr.16: 89971696-89971696 on Build GRCh38
4541126 c.294G>A p.L98L Chr.16: 89971629-89971629 on Build GRCh38
3378039 c.135G>T p.K45N Chr.16: 89971788-89971788 on Build GRCh38
7127864 c.108G>A p.A36A Chr.16: 89971815-89971815 on Build GRCh38
7269047 c.330C>T p.A110A Chr.16: 89971593-89971593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766918940 Chr.16: 89971676-89971676 on Build GRCh38
rs111799449 Chr.16: 89971728-89971728 on Build GRCh38
rs748182989 Chr.16: 89971756-89971756 on Build GRCh38
rs927105371 Chr.16: 89972125-89972125 on Build GRCh38
rs777571853 Chr.16: 89971617-89971617 on Build GRCh38
rs151289005 Chr.16: 89971820-89971821 on Build GRCh38
rs933774293 Chr.16: 89971761-89971761 on Build GRCh38
rs1026319388 Chr.16: 89971908-89971908 on Build GRCh38
rs774546325 Chr.16: 89971839-89971839 on Build GRCh38
rs772730490 Chr.16: 89971849-89971849 on Build GRCh38
rs371786344 Chr.16: 89971833-89971833 on Build GRCh38
rs368302561 Chr.16: 89971783-89971783 on Build GRCh38
rs747000696 Chr.16: 89971797-89971797 on Build GRCh38
rs985190195 Chr.16: 89972081-89972081 on Build GRCh38
rs369610866 Chr.16: 89971897-89971897 on Build GRCh38
rs952236628 Chr.16: 89971702-89971703 on Build GRCh38
rs1006248710 Chr.16: 89971922-89971922 on Build GRCh38
rs752623220 Chr.16: 89971881-89971881 on Build GRCh38
rs775245284 Chr.16: 89971708-89971755 on Build GRCh38
rs769398701 Chr.16: 89971920-89971920 on Build GRCh38
rs776131944 Chr.16: 89971913-89971913 on Build GRCh38
rs368092004 Chr.16: 89971758-89971758 on Build GRCh38
rs772499643 Chr.16: 89971902-89971902 on Build GRCh38
rs561646705 Chr.16: 89972080-89972080 on Build GRCh38
rs766238656 Chr.16: 89971870-89971870 on Build GRCh38
rs937161396 Chr.16: 89972135-89972135 on Build GRCh38
rs775243059 Chr.16: 89971641-89971641 on Build GRCh38
rs760701692 Chr.16: 89971732-89971733 on Build GRCh38
rs36114385 Chr.16: 89971961-89971961 on Build GRCh38
rs369550259 Chr.16: 89971607-89971607 on Build GRCh38
rs199928868 Chr.16: 89971790-89971790 on Build GRCh38
rs745835076 Chr.16: 89971635-89971635 on Build GRCh38
rs761371246 Chr.16: 89971931-89971931 on Build GRCh38
rs138681098 Chr.16: 89971731-89971731 on Build GRCh38
rs768785199 Chr.16: 89971744-89971744 on Build GRCh38
rs756779724 Chr.16: 89971775-89971775 on Build GRCh38
rs191742888 Chr.16: 89971982-89971982 on Build GRCh38
rs763683293 Chr.16: 89971705-89971705 on Build GRCh38
rs535797532 Chr.16: 89971894-89971894 on Build GRCh38
rs923436059 Chr.16: 89972052-89972052 on Build GRCh38
rs748942338 Chr.16: 89971620-89971620 on Build GRCh38
rs763175342 Chr.16: 89971662-89971662 on Build GRCh38
rs879850437 Chr.16: 89971661-89971661 on Build GRCh38
rs946367337 Chr.16: 89972096-89972096 on Build GRCh38
rs763375953 Chr.16: 89971760-89971760 on Build GRCh38
rs755752023 Chr.16: 89971693-89971693 on Build GRCh38
rs760826828 Chr.16: 89971644-89971644 on Build GRCh38
rs954961931 Chr.16: 89972027-89972027 on Build GRCh38
rs761254363 Chr.16: 89971916-89971916 on Build GRCh38
rs1042843740 Chr.16: 89971835-89971835 on Build GRCh38
rs770905097 Chr.16: 89971627-89971627 on Build GRCh38
rs774891030 Chr.16: 89972006-89972006 on Build GRCh38
rs373124572 Chr.16: 89971900-89971900 on Build GRCh38
rs952042869 Chr.16: 89971917-89971917 on Build GRCh38
rs746248312 Chr.16: 89971901-89971901 on Build GRCh38
rs373716402 Chr.16: 89971654-89971654 on Build GRCh38
rs781108345 Chr.16: 89971880-89971880 on Build GRCh38
rs767285985 Chr.16: 89971971-89971971 on Build GRCh38
rs765628611 Chr.16: 89971648-89971648 on Build GRCh38
rs372741187 Chr.16: 89971828-89971828 on Build GRCh38
rs756941255 Chr.16: 89971709-89971709 on Build GRCh38
rs909397702 Chr.16: 89971685-89971685 on Build GRCh38
rs988414192 Chr.16: 89972061-89972061 on Build GRCh38
rs762494309 Chr.16: 89971867-89971867 on Build GRCh38
rs759941453 Chr.16: 89971840-89971840 on Build GRCh38
rs751943762 Chr.16: 89971691-89971691 on Build GRCh38
rs756273651 Chr.16: 89971890-89971890 on Build GRCh38
rs910864486 Chr.16: 89972095-89972095 on Build GRCh38
rs13334308 Chr.16: 89972114-89972114 on Build GRCh38
rs1042772058 Chr.16: 89972124-89972124 on Build GRCh38
rs201646567 Chr.16: 89971739-89971739 on Build GRCh38
rs947084279 Chr.16: 89972123-89972123 on Build GRCh38
rs533109691 Chr.16: 89971639-89971639 on Build GRCh38
rs565751252 Chr.16: 89971634-89971634 on Build GRCh38
rs375347228 Chr.16: 89971781-89971781 on Build GRCh38
rs759390317 Chr.16: 89971963-89971963 on Build GRCh38
rs779703362 Chr.16: 89971793-89971793 on Build GRCh38
rs182290933 Chr.16: 89971930-89971930 on Build GRCh38
rs573546633 Chr.16: 89972112-89972112 on Build GRCh38
rs186944870 Chr.16: 89971953-89971953 on Build GRCh38
rs764653710 Chr.16: 89971862-89971862 on Build GRCh38
rs781055454 Chr.16: 89971813-89971813 on Build GRCh38
rs754937559 Chr.16: 89971805-89971805 on Build GRCh38
rs778670464 Chr.16: 89971723-89971723 on Build GRCh38
rs965223399 Chr.16: 89971979-89971979 on Build GRCh38
rs377274836 Chr.16: 89971815-89971815 on Build GRCh38
rs367596729 Chr.16: 89971829-89971829 on Build GRCh38
rs543478396 Chr.16: 89972077-89972077 on Build GRCh38
rs769572214 Chr.16: 89971771-89971771 on Build GRCh38
rs572562810 Chr.16: 89971910-89971910 on Build GRCh38
rs758071606 Chr.16: 89971738-89971738 on Build GRCh38
rs754707627 Chr.16: 89971873-89971873 on Build GRCh38
rs761413813 Chr.16: 89971861-89971861 on Build GRCh38
rs145823571 Chr.16: 89971706-89971706 on Build GRCh38
rs554415314 Chr.16: 89971895-89971895 on Build GRCh38
rs751375340 Chr.16: 89971871-89971871 on Build GRCh38
rs370787484 Chr.16: 89971764-89971764 on Build GRCh38
rs760096518 Chr.16: 89971772-89971772 on Build GRCh38
rs776908625 Chr.16: 89971647-89971647 on Build GRCh38
rs974973666 Chr.16: 89971988-89971988 on Build GRCh38
rs770022100 Chr.16: 89971757-89971757 on Build GRCh38
rs370186771 Chr.16: 89971736-89971736 on Build GRCh38
rs750152430 Chr.16: 89971866-89971866 on Build GRCh38
rs776084124 Chr.16: 89971860-89971860 on Build GRCh38
rs776672956 Chr.16: 89971748-89971748 on Build GRCh38
rs753430908 Chr.16: 89971773-89971773 on Build GRCh38

View Full Product Details