Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40898335-40898811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2G1P
Entrez Gene ID
22952
Gene Name
cytochrome P450 family 2 subfamily G member 1, pseudogene
Gene Aliases
CYP2G1, CYP2G2P, CYP2GP1
UniGene
Hs.738699
Gene Chromosome Location
Chr.19: 40890826-40900508 on Build GRCh38
Transcripts
NR_040249.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5897171 c.254C>T p.S85F Chr.19: 40898573-40898573 on Build GRCh38
4773719 c.126C>T p.D42D Chr.19: 40898445-40898445 on Build GRCh38
6873097 c.178C>T p.P60S Chr.19: 40898497-40898497 on Build GRCh38
6898019 c.204T>C p.F68F Chr.19: 40898523-40898523 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs940885022 Chr.19: 40898664-40898664 on Build GRCh38
rs114174233 Chr.19: 40898671-40898671 on Build GRCh38
rs967241617 Chr.19: 40898505-40898505 on Build GRCh38
rs184417744 Chr.19: 40898483-40898483 on Build GRCh38
rs559896605 Chr.19: 40898536-40898536 on Build GRCh38
rs866421576 Chr.19: 40898690-40898690 on Build GRCh38
rs1049608668 Chr.19: 40898420-40898420 on Build GRCh38
rs190506366 Chr.19: 40898491-40898491 on Build GRCh38
rs201365230 Chr.19: 40898445-40898445 on Build GRCh38
rs887983804 Chr.19: 40898421-40898421 on Build GRCh38
rs200739912 Chr.19: 40898623-40898623 on Build GRCh38
rs902003742 Chr.19: 40898708-40898708 on Build GRCh38
rs1021760809 Chr.19: 40898499-40898499 on Build GRCh38
rs758708657 Chr.19: 40898422-40898422 on Build GRCh38
rs959973781 Chr.19: 40898546-40898546 on Build GRCh38
rs977634493 Chr.19: 40898515-40898515 on Build GRCh38
rs758749874 Chr.19: 40898449-40898449 on Build GRCh38
rs182283491 Chr.19: 40898539-40898539 on Build GRCh38
rs984899403 Chr.19: 40898653-40898653 on Build GRCh38
rs1028642342 Chr.19: 40898478-40898478 on Build GRCh38
rs771848734 Chr.19: 40898628-40898628 on Build GRCh38
rs997712331 Chr.19: 40898465-40898465 on Build GRCh38
rs780447497 Chr.19: 40898476-40898476 on Build GRCh38
rs556127089 Chr.19: 40898407-40898407 on Build GRCh38
rs149738723 Chr.19: 40898602-40898602 on Build GRCh38
rs765675206 Chr.19: 40898439-40898440 on Build GRCh38
rs948140657 Chr.19: 40898377-40898377 on Build GRCh38
rs538077538 Chr.19: 40898446-40898446 on Build GRCh38
rs751558720 Chr.19: 40898759-40898759 on Build GRCh38
rs770582549 Chr.19: 40898583-40898583 on Build GRCh38
rs544299943 Chr.19: 40898479-40898479 on Build GRCh38
rs140730836 Chr.19: 40898492-40898492 on Build GRCh38
rs909247645 Chr.19: 40898663-40898663 on Build GRCh38
rs1036616384 Chr.19: 40898454-40898454 on Build GRCh38
rs201010762 Chr.19: 40898751-40898751 on Build GRCh38
rs747334428 Chr.19: 40898456-40898456 on Build GRCh38
rs531288738 Chr.19: 40898634-40898634 on Build GRCh38
rs3852871 Chr.19: 40898629-40898629 on Build GRCh38
rs369806971 Chr.19: 40898780-40898780 on Build GRCh38

View Full Product Details