Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 132257684-132258233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD30BL
Entrez Gene ID
554226
Gene Name
ankyrin repeat domain 30B-like
Gene Aliases
ANKRD30BP3, NCRNA00164
UniGene
Hs.380689
Gene Chromosome Location
Chr.2: 132147591-132257969 on Build GRCh38
Transcripts
NR_027020.2

SNP Detail

SNP ID Chromosome Location
rs796770891 Chr.2: 132258155-132258155 on Build GRCh38
rs547660856 Chr.2: 132257917-132257917 on Build GRCh38
rs75269596 Chr.2: 132257967-132257967 on Build GRCh38
rs11677717 Chr.2: 132257930-132257930 on Build GRCh38
rs371433622 Chr.2: 132258080-132258080 on Build GRCh38
rs76386964 Chr.2: 132257835-132257835 on Build GRCh38
rs967110047 Chr.2: 132258131-132258132 on Build GRCh38
rs79590355 Chr.2: 132257749-132257749 on Build GRCh38
rs78724730 Chr.2: 132257934-132257934 on Build GRCh38
rs141411231 Chr.2: 132258121-132258121 on Build GRCh38
rs71986979 Chr.2: 132257943-132257944 on Build GRCh38
rs10175641 Chr.2: 132258003-132258003 on Build GRCh38
rs796094460 Chr.2: 132257891-132257891 on Build GRCh38
rs80347967 Chr.2: 132258071-132258071 on Build GRCh38
rs796392177 Chr.2: 132258040-132258040 on Build GRCh38
rs772794619 Chr.2: 132258098-132258098 on Build GRCh38
rs78481177 Chr.2: 132257954-132257954 on Build GRCh38
rs965962185 Chr.2: 132257751-132257751 on Build GRCh38
rs75143443 Chr.2: 132258196-132258196 on Build GRCh38
rs191218586 Chr.2: 132258031-132258031 on Build GRCh38
rs79930168 Chr.2: 132258077-132258077 on Build GRCh38
rs995632412 Chr.2: 132257866-132257866 on Build GRCh38
rs145538491 Chr.2: 132257846-132257846 on Build GRCh38
rs111389075 Chr.2: 132258052-132258052 on Build GRCh38
rs77919965 Chr.2: 132257736-132257736 on Build GRCh38
rs966358592 Chr.2: 132257902-132257902 on Build GRCh38
rs73957388 Chr.2: 132258193-132258193 on Build GRCh38
rs150256471 Chr.2: 132258148-132258148 on Build GRCh38
rs563824880 Chr.2: 132258146-132258146 on Build GRCh38
rs796150878 Chr.2: 132257775-132257775 on Build GRCh38
rs112876939 Chr.2: 132258100-132258100 on Build GRCh38
rs796534496 Chr.2: 132257723-132257723 on Build GRCh38
rs112494139 Chr.2: 132257801-132257801 on Build GRCh38
rs797001893 Chr.2: 132257773-132257773 on Build GRCh38
rs74890607 Chr.2: 132258053-132258053 on Build GRCh38
rs36192781 Chr.2: 132257937-132257937 on Build GRCh38
rs74753654 Chr.2: 132257858-132257858 on Build GRCh38
rs79987838 Chr.2: 132258009-132258009 on Build GRCh38
rs76632898 Chr.2: 132258093-132258093 on Build GRCh38
rs78892339 Chr.2: 132258171-132258171 on Build GRCh38
rs141595911 Chr.2: 132257868-132257868 on Build GRCh38
rs796111826 Chr.2: 132258042-132258043 on Build GRCh38
rs199810985 Chr.2: 132257863-132257863 on Build GRCh38
rs113794568 Chr.2: 132257795-132257795 on Build GRCh38
rs796071882 Chr.2: 132257705-132257705 on Build GRCh38
rs977168129 Chr.2: 132257800-132257800 on Build GRCh38
rs548265569 Chr.2: 132257809-132257809 on Build GRCh38
rs578757 Chr.2: 132257737-132257737 on Build GRCh38
rs34704394 Chr.2: 132258208-132258208 on Build GRCh38
rs75036976 Chr.2: 132257720-132257720 on Build GRCh38
rs144358225 Chr.2: 132257876-132257876 on Build GRCh38
rs373482090 Chr.2: 132258041-132258041 on Build GRCh38
rs796266908 Chr.2: 132257804-132257804 on Build GRCh38
rs73957387 Chr.2: 132258143-132258143 on Build GRCh38
rs75945497 Chr.2: 132257712-132257712 on Build GRCh38
rs139828151 Chr.2: 132257887-132257887 on Build GRCh38
rs1018757799 Chr.2: 132257897-132257897 on Build GRCh38
rs146652116 Chr.2: 132257901-132257901 on Build GRCh38
rs1056050408 Chr.2: 132257979-132257979 on Build GRCh38
rs931289159 Chr.2: 132257996-132257996 on Build GRCh38
rs113306582 Chr.2: 132258141-132258141 on Build GRCh38
rs150380577 Chr.2: 132257826-132257826 on Build GRCh38
rs147902215 Chr.2: 132257820-132257820 on Build GRCh38
rs111788478 Chr.2: 132257952-132257952 on Build GRCh38
rs765849118 Chr.2: 132257957-132257957 on Build GRCh38
rs985072191 Chr.2: 132258085-132258085 on Build GRCh38
rs112315337 Chr.2: 132257741-132257741 on Build GRCh38
rs869199689 Chr.2: 132257948-132257949 on Build GRCh38
rs796066618 Chr.2: 132258136-132258136 on Build GRCh38
rs113675918 Chr.2: 132258030-132258030 on Build GRCh38
rs76173807 Chr.2: 132257854-132257854 on Build GRCh38
rs78263260 Chr.2: 132258153-132258153 on Build GRCh38
rs796632363 Chr.2: 132257913-132257913 on Build GRCh38
rs762273890 Chr.2: 132257949-132257949 on Build GRCh38
rs1026727912 Chr.2: 132257760-132257760 on Build GRCh38
rs78075547 Chr.2: 132258027-132258027 on Build GRCh38
rs141716220 Chr.2: 132257836-132257836 on Build GRCh38
rs1028846730 Chr.2: 132258033-132258033 on Build GRCh38
rs796109376 Chr.2: 132257792-132257792 on Build GRCh38
rs77397179 Chr.2: 132258150-132258150 on Build GRCh38
rs558226553 Chr.2: 132258016-132258016 on Build GRCh38
rs148060042 Chr.2: 132257859-132257859 on Build GRCh38
rs370616478 Chr.2: 132258005-132258005 on Build GRCh38
rs796477024 Chr.2: 132258010-132258010 on Build GRCh38
rs76329081 Chr.2: 132257825-132257825 on Build GRCh38
rs796726269 Chr.2: 132257846-132257847 on Build GRCh38
rs74212894 Chr.2: 132257948-132257948 on Build GRCh38
rs796266546 Chr.2: 132258203-132258203 on Build GRCh38
rs536388923 Chr.2: 132257986-132257986 on Build GRCh38
rs944152330 Chr.2: 132258208-132258208 on Build GRCh38
rs796490388 Chr.2: 132258209-132258209 on Build GRCh38
rs74673702 Chr.2: 132258135-132258135 on Build GRCh38
rs72869480 Chr.2: 132258046-132258046 on Build GRCh38
rs796800930 Chr.2: 132258004-132258004 on Build GRCh38
rs75962996 Chr.2: 132257830-132257830 on Build GRCh38
rs75692539 Chr.2: 132257729-132257729 on Build GRCh38
rs77561625 Chr.2: 132257904-132257904 on Build GRCh38
rs36148278 Chr.2: 132258028-132258028 on Build GRCh38
rs76775552 Chr.2: 132258200-132258200 on Build GRCh38
rs78690202 Chr.2: 132257790-132257790 on Build GRCh38
rs150562282 Chr.2: 132257997-132257997 on Build GRCh38
rs79130446 Chr.2: 132257945-132257945 on Build GRCh38
rs36139527 Chr.2: 132257921-132257921 on Build GRCh38
rs1056210038 Chr.2: 132257853-132257853 on Build GRCh38
rs982034638 Chr.2: 132257955-132257955 on Build GRCh38
rs928787757 Chr.2: 132257968-132257968 on Build GRCh38
rs148180857 Chr.2: 132257860-132257860 on Build GRCh38
rs72869479 Chr.2: 132258039-132258039 on Build GRCh38
rs1040617592 Chr.2: 132258026-132258026 on Build GRCh38
rs75682799 Chr.2: 132258198-132258198 on Build GRCh38
rs373781359 Chr.2: 132258104-132258104 on Build GRCh38
rs796432939 Chr.2: 132258054-132258055 on Build GRCh38
rs541983518 Chr.2: 132258108-132258108 on Build GRCh38
rs372477900 Chr.2: 132257942-132257943 on Build GRCh38
rs79554046 Chr.2: 132257726-132257726 on Build GRCh38
rs957126192 Chr.2: 132257768-132257768 on Build GRCh38
rs937626056 Chr.2: 132257970-132257970 on Build GRCh38
rs79331692 Chr.2: 132257960-132257960 on Build GRCh38
rs143018696 Chr.2: 132258095-132258095 on Build GRCh38
rs878966179 Chr.2: 132258082-132258085 on Build GRCh38
rs35746979 Chr.2: 132257755-132257756 on Build GRCh38
rs796637160 Chr.2: 132257797-132257797 on Build GRCh38
rs78726101 Chr.2: 132257748-132257748 on Build GRCh38
rs142462538 Chr.2: 132257877-132257878 on Build GRCh38
rs796316743 Chr.2: 132258178-132258178 on Build GRCh38
rs1002141650 Chr.2: 132258070-132258070 on Build GRCh38
rs75705793 Chr.2: 132258076-132258076 on Build GRCh38
rs530179250 Chr.2: 132257778-132257778 on Build GRCh38
rs10172962 Chr.2: 132257841-132257841 on Build GRCh38
rs772160620 Chr.2: 132257932-132257932 on Build GRCh38
rs75932541 Chr.2: 132257939-132257939 on Build GRCh38
rs571529578 Chr.2: 132257870-132257870 on Build GRCh38
rs796944572 Chr.2: 132258065-132258065 on Build GRCh38
rs541235639 Chr.2: 132257710-132257710 on Build GRCh38
rs796665983 Chr.2: 132257851-132257851 on Build GRCh38
rs76066906 Chr.2: 132258192-132258192 on Build GRCh38
rs796841668 Chr.2: 132257787-132257787 on Build GRCh38
rs554628019 Chr.2: 132257998-132257998 on Build GRCh38
rs75625918 Chr.2: 132258036-132258036 on Build GRCh38
rs386391241 Chr.2: 132257944-132257945 on Build GRCh38
rs112524906 Chr.2: 132258057-132258057 on Build GRCh38
rs79256679 Chr.2: 132257730-132257730 on Build GRCh38
rs773021908 Chr.2: 132257811-132257811 on Build GRCh38
rs79736878 Chr.2: 132257985-132257985 on Build GRCh38
rs111563229 Chr.2: 132257951-132257951 on Build GRCh38
rs80035394 Chr.2: 132257719-132257719 on Build GRCh38
rs796919171 Chr.2: 132258134-132258134 on Build GRCh38
rs74693824 Chr.2: 132258055-132258055 on Build GRCh38
rs796947182 Chr.2: 132257867-132257867 on Build GRCh38
rs201258639 Chr.2: 132258175-132258175 on Build GRCh38
rs746267056 Chr.2: 132258019-132258019 on Build GRCh38
rs399181 Chr.2: 132257758-132257758 on Build GRCh38
rs796282813 Chr.2: 132258032-132258032 on Build GRCh38
rs78398186 Chr.2: 132258166-132258166 on Build GRCh38
rs79301155 Chr.2: 132257881-132257881 on Build GRCh38
rs937570131 Chr.2: 132257852-132257853 on Build GRCh38
rs796366340 Chr.2: 132257807-132257807 on Build GRCh38
rs944096330 Chr.2: 132258025-132258025 on Build GRCh38
rs912968494 Chr.2: 132257942-132257942 on Build GRCh38
rs111452280 Chr.2: 132258110-132258110 on Build GRCh38
rs757962982 Chr.2: 132257956-132257956 on Build GRCh38
rs56740690 Chr.2: 132257847-132257847 on Build GRCh38
rs111401445 Chr.2: 132257752-132257752 on Build GRCh38
rs112040002 Chr.2: 132258169-132258169 on Build GRCh38
rs76980088 Chr.2: 132258154-132258154 on Build GRCh38
rs111836108 Chr.2: 132258001-132258001 on Build GRCh38
rs72869482 Chr.2: 132258133-132258133 on Build GRCh38
rs879080052 Chr.2: 132257761-132257761 on Build GRCh38
rs796848146 Chr.2: 132257983-132257983 on Build GRCh38
rs78870510 Chr.2: 132258147-132258147 on Build GRCh38
rs80267931 Chr.2: 132257971-132257971 on Build GRCh38
rs774557423 Chr.2: 132257912-132257912 on Build GRCh38
rs796734057 Chr.2: 132257781-132257781 on Build GRCh38
rs892831071 Chr.2: 132258061-132258061 on Build GRCh38
rs113413216 Chr.2: 132258182-132258182 on Build GRCh38
rs796657870 Chr.2: 132258012-132258012 on Build GRCh38
rs796417884 Chr.2: 132257766-132257766 on Build GRCh38
rs376221501 Chr.2: 132257974-132257974 on Build GRCh38
rs144614601 Chr.2: 132258075-132258075 on Build GRCh38
rs449977 Chr.2: 132257861-132257861 on Build GRCh38
rs77267587 Chr.2: 132258011-132258011 on Build GRCh38
rs80252383 Chr.2: 132257828-132257828 on Build GRCh38
rs139986343 Chr.2: 132257827-132257828 on Build GRCh38
rs76610352 Chr.2: 132257837-132257837 on Build GRCh38
rs989423513 Chr.2: 132257880-132257880 on Build GRCh38
rs202110773 Chr.2: 132257780-132257780 on Build GRCh38
rs1019882508 Chr.2: 132258112-132258112 on Build GRCh38
rs530872076 Chr.2: 132258191-132258191 on Build GRCh38
rs554205 Chr.2: 132258115-132258115 on Build GRCh38
rs546185684 Chr.2: 132258202-132258202 on Build GRCh38
rs796211982 Chr.2: 132257976-132257976 on Build GRCh38
rs376376834 Chr.2: 132258078-132258078 on Build GRCh38
rs188125715 Chr.2: 132258023-132258023 on Build GRCh38
rs437455 Chr.2: 132257903-132257903 on Build GRCh38
rs77644836 Chr.2: 132257946-132257946 on Build GRCh38
rs538891194 Chr.2: 132257892-132257892 on Build GRCh38
rs200631194 Chr.2: 132258149-132258151 on Build GRCh38
rs958883818 Chr.2: 132258170-132258170 on Build GRCh38
rs530909948 Chr.2: 132257849-132257849 on Build GRCh38
rs921575425 Chr.2: 132257765-132257765 on Build GRCh38
rs796519735 Chr.2: 132257858-132257859 on Build GRCh38
rs78844169 Chr.2: 132257812-132257812 on Build GRCh38
rs796533755 Chr.2: 132257988-132257988 on Build GRCh38
rs974830827 Chr.2: 132257754-132257754 on Build GRCh38
rs541321348 Chr.2: 132258029-132258029 on Build GRCh38
rs36160636 Chr.2: 132257864-132257864 on Build GRCh38
rs796531085 Chr.2: 132257884-132257884 on Build GRCh38
rs553188810 Chr.2: 132258044-132258044 on Build GRCh38
rs796678963 Chr.2: 132257802-132257802 on Build GRCh38
rs201079734 Chr.2: 132257703-132257703 on Build GRCh38
rs36147689 Chr.2: 132258091-132258091 on Build GRCh38
rs72869481 Chr.2: 132258064-132258064 on Build GRCh38
rs75473717 Chr.2: 132257829-132257829 on Build GRCh38
rs79116859 Chr.2: 132257727-132257727 on Build GRCh38

View Full Product Details