Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130098688-130099244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM86HP
Entrez Gene ID
729375
Gene Name
family with sequence similarity 86, member A pseudogene
Gene Aliases
-
UniGene
Hs.123633
Gene Chromosome Location
Chr.3: 130097782-130111433 on Build GRCh38
Transcripts
NR_024252.1

SNP Detail

SNP ID Chromosome Location
rs58288206 Chr.3: 130098924-130098924 on Build GRCh38
rs559852419 Chr.3: 130099154-130099154 on Build GRCh38
rs1057205936 Chr.3: 130098826-130098826 on Build GRCh38
rs1019116496 Chr.3: 130099186-130099186 on Build GRCh38
rs75003290 Chr.3: 130098742-130098742 on Build GRCh38
rs6788466 Chr.3: 130098836-130098836 on Build GRCh38
rs6764369 Chr.3: 130098880-130098880 on Build GRCh38
rs533273761 Chr.3: 130099106-130099106 on Build GRCh38
rs1023307494 Chr.3: 130098866-130098866 on Build GRCh38
rs745774697 Chr.3: 130099159-130099159 on Build GRCh38
rs563199710 Chr.3: 130098752-130098752 on Build GRCh38
rs868447509 Chr.3: 130098861-130098861 on Build GRCh38
rs202210309 Chr.3: 130098757-130098757 on Build GRCh38
rs994705746 Chr.3: 130098897-130098897 on Build GRCh38
rs540329366 Chr.3: 130098979-130098979 on Build GRCh38
rs183706721 Chr.3: 130099045-130099045 on Build GRCh38
rs551384023 Chr.3: 130099080-130099080 on Build GRCh38
rs186008777 Chr.3: 130098832-130098832 on Build GRCh38
rs947654849 Chr.3: 130099003-130099003 on Build GRCh38
rs1047608466 Chr.3: 130099170-130099170 on Build GRCh38
rs761181382 Chr.3: 130099158-130099158 on Build GRCh38
rs201387340 Chr.3: 130099101-130099101 on Build GRCh38
rs987194003 Chr.3: 130098928-130098928 on Build GRCh38
rs948737758 Chr.3: 130099100-130099100 on Build GRCh38
rs868070317 Chr.3: 130098784-130098784 on Build GRCh38
rs886291614 Chr.3: 130099171-130099171 on Build GRCh38
rs549069980 Chr.3: 130098833-130098833 on Build GRCh38
rs201142223 Chr.3: 130098924-130098924 on Build GRCh38
rs567301307 Chr.3: 130098834-130098834 on Build GRCh38
rs191173802 Chr.3: 130098937-130098937 on Build GRCh38
rs751171007 Chr.3: 130098809-130098809 on Build GRCh38
rs2587971 Chr.3: 130098733-130098733 on Build GRCh38
rs577550539 Chr.3: 130098885-130098885 on Build GRCh38
rs760977129 Chr.3: 130098923-130098924 on Build GRCh38
rs562216971 Chr.3: 130099001-130099001 on Build GRCh38
rs967691142 Chr.3: 130099212-130099213 on Build GRCh38
rs976087722 Chr.3: 130098759-130098759 on Build GRCh38
rs1021982981 Chr.3: 130099211-130099211 on Build GRCh38
rs2587970 Chr.3: 130098961-130098961 on Build GRCh38
rs899169074 Chr.3: 130099126-130099126 on Build GRCh38
rs996324512 Chr.3: 130099206-130099206 on Build GRCh38
rs563150597 Chr.3: 130099055-130099055 on Build GRCh38
rs200352094 Chr.3: 130098977-130098977 on Build GRCh38
rs903346909 Chr.3: 130098790-130098790 on Build GRCh38
rs533713934 Chr.3: 130099077-130099077 on Build GRCh38
rs934790214 Chr.3: 130099049-130099049 on Build GRCh38
rs1000184469 Chr.3: 130098900-130098900 on Build GRCh38
rs1043278676 Chr.3: 130098787-130098787 on Build GRCh38
rs764295418 Chr.3: 130098917-130098917 on Build GRCh38
rs778942548 Chr.3: 130099062-130099062 on Build GRCh38
rs6788479 Chr.3: 130098865-130098865 on Build GRCh38
rs138565622 Chr.3: 130098725-130098725 on Build GRCh38
rs566566639 Chr.3: 130098781-130098781 on Build GRCh38
rs538065173 Chr.3: 130098863-130098863 on Build GRCh38
rs527281677 Chr.3: 130099173-130099173 on Build GRCh38
rs200253006 Chr.3: 130098922-130098922 on Build GRCh38
rs1057467616 Chr.3: 130099079-130099079 on Build GRCh38
rs758249920 Chr.3: 130099193-130099193 on Build GRCh38
rs934757850 Chr.3: 130098802-130098802 on Build GRCh38
rs867512541 Chr.3: 130099212-130099212 on Build GRCh38
rs539426067 Chr.3: 130098915-130098915 on Build GRCh38
rs978987841 Chr.3: 130099008-130099008 on Build GRCh38
rs917302508 Chr.3: 130099082-130099082 on Build GRCh38
rs551422360 Chr.3: 130098760-130098760 on Build GRCh38
rs548775426 Chr.3: 130099176-130099176 on Build GRCh38
rs964524845 Chr.3: 130099191-130099191 on Build GRCh38

View Full Product Details