Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 198080126-198080688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD18DP
Entrez Gene ID
348840
Gene Name
ankyrin repeat domain 18D, pseudogene
Gene Aliases
-
UniGene
Hs.512227
Gene Chromosome Location
Chr.3: 198057531-198080671 on Build GRCh38
Transcripts
NR_003291.1

SNP Detail

SNP ID Chromosome Location
rs527556211 Chr.3: 198080496-198080496 on Build GRCh38
rs534090160 Chr.3: 198080194-198080194 on Build GRCh38
rs143389077 Chr.3: 198080541-198080541 on Build GRCh38
rs1043988993 Chr.3: 198080297-198080297 on Build GRCh38
rs1009285899 Chr.3: 198080276-198080276 on Build GRCh38
rs373125975 Chr.3: 198080449-198080449 on Build GRCh38
rs114314433 Chr.3: 198080425-198080425 on Build GRCh38
rs926300221 Chr.3: 198080151-198080151 on Build GRCh38
rs892212474 Chr.3: 198080254-198080254 on Build GRCh38
rs980378685 Chr.3: 198080395-198080395 on Build GRCh38
rs997490001 Chr.3: 198080329-198080329 on Build GRCh38
rs928891629 Chr.3: 198080416-198080416 on Build GRCh38
rs149469977 Chr.3: 198080306-198080306 on Build GRCh38
rs939012676 Chr.3: 198080189-198080189 on Build GRCh38
rs950745020 Chr.3: 198080524-198080524 on Build GRCh38
rs538069591 Chr.3: 198080596-198080596 on Build GRCh38
rs1026766484 Chr.3: 198080508-198080508 on Build GRCh38
rs147648028 Chr.3: 198080285-198080285 on Build GRCh38
rs1026272977 Chr.3: 198080338-198080338 on Build GRCh38
rs561342420 Chr.3: 198080340-198080340 on Build GRCh38
rs970231930 Chr.3: 198080388-198080388 on Build GRCh38
rs550239575 Chr.3: 198080439-198080439 on Build GRCh38
rs566467471 Chr.3: 198080473-198080473 on Build GRCh38
rs547836982 Chr.3: 198080472-198080472 on Build GRCh38
rs909409642 Chr.3: 198080386-198080386 on Build GRCh38
rs542704037 Chr.3: 198080332-198080332 on Build GRCh38
rs1017463379 Chr.3: 198080316-198080316 on Build GRCh38
rs950867940 Chr.3: 198080362-198080362 on Build GRCh38
rs529682065 Chr.3: 198080462-198080462 on Build GRCh38
rs373950810 Chr.3: 198080261-198080261 on Build GRCh38
rs1038817670 Chr.3: 198080476-198080476 on Build GRCh38
rs137883806 Chr.3: 198080242-198080242 on Build GRCh38
rs116400810 Chr.3: 198080598-198080598 on Build GRCh38
rs1056149393 Chr.3: 198080426-198080426 on Build GRCh38
rs570647816 Chr.3: 198080543-198080543 on Build GRCh38
rs369853773 Chr.3: 198080328-198080328 on Build GRCh38
rs183177377 Chr.3: 198080179-198080179 on Build GRCh38
rs928754423 Chr.3: 198080669-198080669 on Build GRCh38
rs758364462 Chr.3: 198080480-198080480 on Build GRCh38
rs13062943 Chr.3: 198080370-198080370 on Build GRCh38
rs73891889 Chr.3: 198080433-198080433 on Build GRCh38
rs1007456826 Chr.3: 198080310-198080310 on Build GRCh38
rs558740428 Chr.3: 198080226-198080226 on Build GRCh38
rs901768223 Chr.3: 198080491-198080491 on Build GRCh38
rs376082286 Chr.3: 198080511-198080511 on Build GRCh38
rs9812611 Chr.3: 198080324-198080324 on Build GRCh38
rs1016336397 Chr.3: 198080553-198080553 on Build GRCh38
rs528053172 Chr.3: 198080349-198080349 on Build GRCh38

View Full Product Details