Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 118523305-118523855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP170P1
Entrez Gene ID
645455
Gene Name
centrosomal protein 170 pseudogene 1
Gene Aliases
CEP170L, FAM68B, KIAA0470L
UniGene
Hs.408293
Gene Chromosome Location
Chr.4: 118516340-118554204 on Build GRCh38
Transcripts
NR_003135.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5901297 c.79G>A p.E27K Chr.4: 118523435-118523435 on Build GRCh38
6099054 c.39G>T p.E13D Chr.4: 118523395-118523395 on Build GRCh38
3767665 c.60T>C p.D20D Chr.4: 118523416-118523416 on Build GRCh38
5573137 c.50A>G p.Y17C Chr.4: 118523406-118523406 on Build GRCh38
1050349 c.88A>G p.R30G Chr.4: 118523444-118523444 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs891031814 Chr.4: 118523328-118523328 on Build GRCh38
rs778313694 Chr.4: 118523402-118523402 on Build GRCh38
rs916549141 Chr.4: 118523502-118523502 on Build GRCh38
rs935179467 Chr.4: 118523585-118523585 on Build GRCh38
rs781371628 Chr.4: 118523439-118523439 on Build GRCh38
rs566617600 Chr.4: 118523814-118523814 on Build GRCh38
rs779831492 Chr.4: 118523418-118523418 on Build GRCh38
rs533222513 Chr.4: 118523588-118523588 on Build GRCh38
rs886847851 Chr.4: 118523725-118523725 on Build GRCh38
rs1015083428 Chr.4: 118523787-118523787 on Build GRCh38
rs983683089 Chr.4: 118523457-118523457 on Build GRCh38
rs1005045863 Chr.4: 118523785-118523785 on Build GRCh38
rs746800893 Chr.4: 118523422-118523422 on Build GRCh38
rs546501669 Chr.4: 118523658-118523658 on Build GRCh38
rs992599042 Chr.4: 118523474-118523474 on Build GRCh38
rs201496090 Chr.4: 118523460-118523460 on Build GRCh38
rs866669311 Chr.4: 118523544-118523544 on Build GRCh38
rs766720818 Chr.4: 118523357-118523357 on Build GRCh38
rs1005851935 Chr.4: 118523706-118523706 on Build GRCh38
rs536925778 Chr.4: 118523576-118523576 on Build GRCh38
rs763463714 Chr.4: 118523486-118523486 on Build GRCh38
rs1039785877 Chr.4: 118523616-118523616 on Build GRCh38
rs556559881 Chr.4: 118523680-118523680 on Build GRCh38
rs200676170 Chr.4: 118523334-118523334 on Build GRCh38
rs751990163 Chr.4: 118523375-118523375 on Build GRCh38
rs185507101 Chr.4: 118523344-118523344 on Build GRCh38
rs961253897 Chr.4: 118523808-118523808 on Build GRCh38
rs960983992 Chr.4: 118523464-118523464 on Build GRCh38
rs765283580 Chr.4: 118523338-118523338 on Build GRCh38
rs948138039 Chr.4: 118523545-118523545 on Build GRCh38
rs768496468 Chr.4: 118523438-118523438 on Build GRCh38
rs758280233 Chr.4: 118523411-118523411 on Build GRCh38
rs943962054 Chr.4: 118523615-118523615 on Build GRCh38
rs780565990 Chr.4: 118523342-118523342 on Build GRCh38
rs756794113 Chr.4: 118523396-118523396 on Build GRCh38
rs899460955 Chr.4: 118523629-118523629 on Build GRCh38
rs561440033 Chr.4: 118523381-118523381 on Build GRCh38
rs567977490 Chr.4: 118523385-118523385 on Build GRCh38
rs527472723 Chr.4: 118523608-118523608 on Build GRCh38
rs995237460 Chr.4: 118523657-118523657 on Build GRCh38
rs763402656 Chr.4: 118523355-118523355 on Build GRCh38
rs530471626 Chr.4: 118523384-118523384 on Build GRCh38
rs3109965 Chr.4: 118523686-118523686 on Build GRCh38
rs1052249953 Chr.4: 118523607-118523608 on Build GRCh38
rs1024074416 Chr.4: 118523824-118523824 on Build GRCh38
rs769878399 Chr.4: 118523456-118523456 on Build GRCh38
rs572872290 Chr.4: 118523333-118523333 on Build GRCh38
rs564261290 Chr.4: 118523577-118523577 on Build GRCh38
rs750322295 Chr.4: 118523409-118523409 on Build GRCh38
rs775169671 Chr.4: 118523327-118523327 on Build GRCh38
rs1026665106 Chr.4: 118523673-118523673 on Build GRCh38
rs952193342 Chr.4: 118523450-118523450 on Build GRCh38
rs576611687 Chr.4: 118523738-118523738 on Build GRCh38

View Full Product Details