Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 118553615-118554165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP170P1
Entrez Gene ID
645455
Gene Name
centrosomal protein 170 pseudogene 1
Gene Aliases
CEP170L, FAM68B, KIAA0470L
UniGene
Hs.408293
Gene Chromosome Location
Chr.4: 118516340-118554204 on Build GRCh38
Transcripts
NR_003135.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5898839 c.760C>T p.H254Y Chr.4: 118553978-118553978 on Build GRCh38
3409024 c.845C>T p.P282L Chr.4: 118554063-118554063 on Build GRCh38
6626937 c.848A>G p.D283G Chr.4: 118554066-118554066 on Build GRCh38
4671628 c.808_810delGAG p.E270delE Chr.4: 118554026-118554028 on Build GRCh38
6220163 c.624delT p.L209fs*4 Chr.4: 118553842-118553842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990912690 Chr.4: 118553879-118553879 on Build GRCh38
rs758146469 Chr.4: 118554115-118554115 on Build GRCh38
rs780782141 Chr.4: 118553802-118553802 on Build GRCh38
rs540047767 Chr.4: 118554081-118554081 on Build GRCh38
rs762315258 Chr.4: 118554055-118554055 on Build GRCh38
rs765578437 Chr.4: 118553934-118553934 on Build GRCh38
rs780216028 Chr.4: 118554031-118554031 on Build GRCh38
rs201105783 Chr.4: 118553846-118553846 on Build GRCh38
rs553248589 Chr.4: 118554123-118554123 on Build GRCh38
rs571025407 Chr.4: 118554018-118554018 on Build GRCh38
rs754601727 Chr.4: 118553801-118553801 on Build GRCh38
rs768762666 Chr.4: 118553900-118553900 on Build GRCh38
rs547441323 Chr.4: 118554020-118554020 on Build GRCh38
rs893648448 Chr.4: 118553643-118553643 on Build GRCh38
rs775745343 Chr.4: 118553895-118553895 on Build GRCh38
rs1050928967 Chr.4: 118553942-118553942 on Build GRCh38
rs762249029 Chr.4: 118553932-118553932 on Build GRCh38
rs768672893 Chr.4: 118554035-118554035 on Build GRCh38
rs755656283 Chr.4: 118553998-118553998 on Build GRCh38
rs1026464956 Chr.4: 118553757-118553757 on Build GRCh38
rs902449995 Chr.4: 118553698-118553698 on Build GRCh38
rs3109971 Chr.4: 118554094-118554094 on Build GRCh38
rs763389248 Chr.4: 118554066-118554066 on Build GRCh38
rs763443044 Chr.4: 118553962-118553962 on Build GRCh38
rs752648411 Chr.4: 118553810-118553810 on Build GRCh38
rs770198656 Chr.4: 118554057-118554057 on Build GRCh38
rs868594257 Chr.4: 118553961-118553961 on Build GRCh38
rs548693281 Chr.4: 118553719-118553719 on Build GRCh38
rs753763436 Chr.4: 118554003-118554003 on Build GRCh38
rs771900933 Chr.4: 118554028-118554028 on Build GRCh38
rs745634748 Chr.4: 118554020-118554020 on Build GRCh38
rs747262288 Chr.4: 118554034-118554034 on Build GRCh38
rs889445864 Chr.4: 118553728-118553728 on Build GRCh38
rs751173778 Chr.4: 118553783-118553783 on Build GRCh38
rs1029774950 Chr.4: 118553718-118553718 on Build GRCh38
rs537423308 Chr.4: 118553776-118553776 on Build GRCh38
rs1042324092 Chr.4: 118553693-118553693 on Build GRCh38
rs752448167 Chr.4: 118553986-118553986 on Build GRCh38
rs752360343 Chr.4: 118554074-118554074 on Build GRCh38
rs750758715 Chr.4: 118553935-118553935 on Build GRCh38
rs772272931 Chr.4: 118553888-118553888 on Build GRCh38
rs998097989 Chr.4: 118553714-118553714 on Build GRCh38
rs749225830 Chr.4: 118553844-118553844 on Build GRCh38
rs1016603664 Chr.4: 118553736-118553736 on Build GRCh38
rs951063845 Chr.4: 118553794-118553794 on Build GRCh38
rs778598211 Chr.4: 118553857-118553857 on Build GRCh38
rs766893792 Chr.4: 118554070-118554070 on Build GRCh38
rs757608222 Chr.4: 118553771-118553771 on Build GRCh38
rs765263834 Chr.4: 118553795-118553795 on Build GRCh38
rs568513664 Chr.4: 118553734-118553734 on Build GRCh38
rs756846571 Chr.4: 118554087-118554087 on Build GRCh38
rs777153621 Chr.4: 118553902-118553902 on Build GRCh38
rs946801442 Chr.4: 118553905-118553905 on Build GRCh38
rs1042640180 Chr.4: 118553918-118553918 on Build GRCh38
rs192196158 Chr.4: 118553999-118553999 on Build GRCh38
rs750412230 Chr.4: 118554110-118554110 on Build GRCh38
rs963526968 Chr.4: 118553741-118553741 on Build GRCh38
rs747174775 Chr.4: 118554139-118554139 on Build GRCh38
rs937957372 Chr.4: 118553816-118553816 on Build GRCh38
rs757142200 Chr.4: 118554016-118554016 on Build GRCh38
rs528537964 Chr.4: 118553710-118553710 on Build GRCh38
rs760429902 Chr.4: 118554080-118554080 on Build GRCh38
rs1011124922 Chr.4: 118553687-118553687 on Build GRCh38
rs765060534 Chr.4: 118554097-118554097 on Build GRCh38
rs973956363 Chr.4: 118553749-118553749 on Build GRCh38
rs559801130 Chr.4: 118553645-118553645 on Build GRCh38
rs889457296 Chr.4: 118554061-118554061 on Build GRCh38
rs755744200 Chr.4: 118553817-118553817 on Build GRCh38
rs767267965 Chr.4: 118553966-118553966 on Build GRCh38
rs773828386 Chr.4: 118554060-118554060 on Build GRCh38
rs777833670 Chr.4: 118553822-118553822 on Build GRCh38
rs760719462 Chr.4: 118553896-118553896 on Build GRCh38
rs746031634 Chr.4: 118553876-118553876 on Build GRCh38
rs776587601 Chr.4: 118554042-118554042 on Build GRCh38

View Full Product Details