Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 12674753-12675262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMPD4
Entrez Gene ID
9758
Gene Name
FERM and PDZ domain containing 4
Gene Aliases
PDZD10, PDZK10
UniGene
Hs.657507
Gene Chromosome Location
Chr.X: 12137329-12724523 on Build GRCh38
Transcripts
NM_014728.3 XM_005274632.3 XM_011545613.2 XM_017029983.1 XM_017029984.1 XM_017029985.1 XM_017029986.1

SNP Detail

SNP ID Chromosome Location
rs911279894 Chr.X: 12674851-12674851 on Build GRCh38
rs750345258 Chr.X: 12675151-12675151 on Build GRCh38
rs956789413 Chr.X: 12674801-12674801 on Build GRCh38
rs190300696 Chr.X: 12674817-12674817 on Build GRCh38
rs113642762 Chr.X: 12674810-12674810 on Build GRCh38
rs896135209 Chr.X: 12675017-12675017 on Build GRCh38
rs748494521 Chr.X: 12674873-12674873 on Build GRCh38
rs749022111 Chr.X: 12675097-12675097 on Build GRCh38
rs764758649 Chr.X: 12675016-12675016 on Build GRCh38
rs898788050 Chr.X: 12674942-12674942 on Build GRCh38
rs1008753955 Chr.X: 12674820-12674820 on Build GRCh38
rs1038905034 Chr.X: 12674891-12674891 on Build GRCh38
rs1023402997 Chr.X: 12675048-12675048 on Build GRCh38
rs369465027 Chr.X: 12674829-12674829 on Build GRCh38
rs899947089 Chr.X: 12674834-12674834 on Build GRCh38
rs997335316 Chr.X: 12675015-12675015 on Build GRCh38
rs749702898 Chr.X: 12674907-12674907 on Build GRCh38
rs768308066 Chr.X: 12675111-12675111 on Build GRCh38
rs755239036 Chr.X: 12674823-12674823 on Build GRCh38
rs769528716 Chr.X: 12675005-12675005 on Build GRCh38
rs113303439 Chr.X: 12675004-12675004 on Build GRCh38
rs993041349 Chr.X: 12675193-12675193 on Build GRCh38
rs773733657 Chr.X: 12674947-12674947 on Build GRCh38
rs773793762 Chr.X: 12674906-12674906 on Build GRCh38
rs758291026 Chr.X: 12675215-12675215 on Build GRCh38
rs376491171 Chr.X: 12674818-12674818 on Build GRCh38
rs1031903049 Chr.X: 12675093-12675093 on Build GRCh38
rs759241185 Chr.X: 12674956-12674956 on Build GRCh38
rs778238913 Chr.X: 12674812-12674812 on Build GRCh38
rs781724060 Chr.X: 12674854-12674854 on Build GRCh38
rs1000700880 Chr.X: 12675139-12675139 on Build GRCh38
rs942882398 Chr.X: 12674889-12674889 on Build GRCh38
rs770483309 Chr.X: 12674933-12674933 on Build GRCh38
rs1017840795 Chr.X: 12674828-12674828 on Build GRCh38
rs183555567 Chr.X: 12674821-12674821 on Build GRCh38
rs770244799 Chr.X: 12674904-12674904 on Build GRCh38
rs186881573 Chr.X: 12675026-12675026 on Build GRCh38
rs771724249 Chr.X: 12674937-12674937 on Build GRCh38
rs913214880 Chr.X: 12674814-12674814 on Build GRCh38

View Full Product Details