Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14042543-14042837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL27RA
Entrez Gene ID
9466
Gene Name
interleukin 27 receptor subunit alpha
Gene Aliases
CRL1, IL-27RA, IL27R, TCCR, WSX1, zcytor1
UniGene
Hs.132781
Gene Chromosome Location
Chr.19: 14031740-14052919 on Build GRCh38
Transcripts
NM_004843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5560362 c.628A>G p.M210V Chr.19: 14042546-14042546 on Build GRCh38
1611655 c.694+3G>A p.? Chr.19: 14042615-14042615 on Build GRCh38
991796 c.626G>A p.R209Q Chr.19: 14042544-14042544 on Build GRCh38
4278481 c.630G>T p.M210I Chr.19: 14042548-14042548 on Build GRCh38
5013383 c.659G>A p.W220* Chr.19: 14042577-14042577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376129161 Chr.19: 14042763-14042763 on Build GRCh38
rs751093690 Chr.19: 14042657-14042657 on Build GRCh38
rs1011273426 Chr.19: 14042681-14042681 on Build GRCh38
rs765199413 Chr.19: 14042632-14042632 on Build GRCh38
rs747735340 Chr.19: 14042566-14042566 on Build GRCh38
rs747764749 Chr.19: 14042697-14042697 on Build GRCh38
rs550622916 Chr.19: 14042605-14042605 on Build GRCh38
rs769827236 Chr.19: 14042744-14042744 on Build GRCh38
rs772990308 Chr.19: 14042573-14042573 on Build GRCh38
rs778410922 Chr.19: 14042637-14042637 on Build GRCh38
rs545007579 Chr.19: 14042665-14042665 on Build GRCh38
rs757904994 Chr.19: 14042707-14042707 on Build GRCh38
rs370468770 Chr.19: 14042804-14042804 on Build GRCh38
rs373161906 Chr.19: 14042638-14042638 on Build GRCh38
rs1029749941 Chr.19: 14042570-14042570 on Build GRCh38
rs745769605 Chr.19: 14042741-14042741 on Build GRCh38
rs375840149 Chr.19: 14042643-14042643 on Build GRCh38
rs374116608 Chr.19: 14042694-14042694 on Build GRCh38
rs149346880 Chr.19: 14042755-14042755 on Build GRCh38
rs551348861 Chr.19: 14042773-14042773 on Build GRCh38
rs777345865 Chr.19: 14042713-14042713 on Build GRCh38
rs139793647 Chr.19: 14042612-14042612 on Build GRCh38
rs764346145 Chr.19: 14042647-14042647 on Build GRCh38
rs200751378 Chr.19: 14042602-14042602 on Build GRCh38
rs757944471 Chr.19: 14042654-14042654 on Build GRCh38
rs369426970 Chr.19: 14042631-14042631 on Build GRCh38
rs750867067 Chr.19: 14042672-14042672 on Build GRCh38
rs202125422 Chr.19: 14042572-14042572 on Build GRCh38
rs937652918 Chr.19: 14042686-14042686 on Build GRCh38
rs775427545 Chr.19: 14042746-14042746 on Build GRCh38
rs1014886094 Chr.19: 14042812-14042812 on Build GRCh38
rs776528868 Chr.19: 14042729-14042729 on Build GRCh38
rs954531939 Chr.19: 14042774-14042774 on Build GRCh38
rs745464064 Chr.19: 14042772-14042772 on Build GRCh38
rs143701993 Chr.19: 14042722-14042722 on Build GRCh38
rs756578619 Chr.19: 14042783-14042783 on Build GRCh38
rs780931987 Chr.19: 14042756-14042756 on Build GRCh38
rs757851653 Chr.19: 14042810-14042810 on Build GRCh38
rs866149927 Chr.19: 14042716-14042716 on Build GRCh38
rs996314559 Chr.19: 14042721-14042721 on Build GRCh38
rs201013102 Chr.19: 14042728-14042728 on Build GRCh38
rs900272318 Chr.19: 14042719-14042719 on Build GRCh38
rs777592553 Chr.19: 14042705-14042705 on Build GRCh38
rs149254897 Chr.19: 14042596-14042596 on Build GRCh38
rs767973999 Chr.19: 14042764-14042764 on Build GRCh38
rs764291163 Chr.19: 14042754-14042754 on Build GRCh38
rs147347872 Chr.19: 14042604-14042604 on Build GRCh38
rs757649812 Chr.19: 14042662-14042662 on Build GRCh38
rs954044298 Chr.19: 14042597-14042597 on Build GRCh38
rs763031588 Chr.19: 14042752-14042752 on Build GRCh38
rs770670251 Chr.19: 14042727-14042727 on Build GRCh38
rs200086680 Chr.19: 14042696-14042696 on Build GRCh38
rs759036408 Chr.19: 14042611-14042611 on Build GRCh38
rs913360433 Chr.19: 14042640-14042640 on Build GRCh38

View Full Product Details