Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108559813-108560171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1524
Entrez Gene ID
57650
Gene Name
KIAA1524
Gene Aliases
CIP2A, p90
UniGene
Hs.591308
Gene Chromosome Location
Chr.3: 108545752-108589644 on Build GRCh38
Transcripts
NM_020890.2 XM_006713716.3 XM_006713717.2 XM_011513056.1 XM_011513057.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6095821 c.1831A>C p.K611Q Chr.3: 108560025-108560025 on Build GRCh38
6410225 c.1909G>T p.E637* Chr.3: 108559861-108559861 on Build GRCh38
6095822 c.1924G>T p.D642Y Chr.3: 108559846-108559846 on Build GRCh38
2916977 c.1948G>A p.A650T Chr.3: 108559822-108559822 on Build GRCh38
4474898 c.1950C>T p.A650A Chr.3: 108559820-108559820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763355650 Chr.3: 108559980-108559983 on Build GRCh38
rs761309478 Chr.3: 108560074-108560074 on Build GRCh38
rs780038413 Chr.3: 108560028-108560028 on Build GRCh38
rs952016984 Chr.3: 108559896-108559896 on Build GRCh38
rs144840000 Chr.3: 108559963-108559963 on Build GRCh38
rs758291203 Chr.3: 108559871-108559871 on Build GRCh38
rs778622391 Chr.3: 108559866-108559866 on Build GRCh38
rs760254281 Chr.3: 108559929-108559929 on Build GRCh38
rs370778599 Chr.3: 108560032-108560032 on Build GRCh38
rs752116940 Chr.3: 108560051-108560051 on Build GRCh38
rs773857938 Chr.3: 108559913-108559913 on Build GRCh38
rs779212663 Chr.3: 108560053-108560053 on Build GRCh38
rs770440194 Chr.3: 108560077-108560077 on Build GRCh38
rs199863584 Chr.3: 108559870-108559870 on Build GRCh38
rs750829979 Chr.3: 108560126-108560128 on Build GRCh38
rs777684967 Chr.3: 108560001-108560001 on Build GRCh38
rs745883711 Chr.3: 108559883-108559883 on Build GRCh38
rs772630260 Chr.3: 108560069-108560069 on Build GRCh38
rs556082866 Chr.3: 108560096-108560096 on Build GRCh38
rs755701187 Chr.3: 108559844-108559844 on Build GRCh38
rs753497319 Chr.3: 108559858-108559858 on Build GRCh38
rs749906882 Chr.3: 108559840-108559840 on Build GRCh38
rs766060866 Chr.3: 108559856-108559856 on Build GRCh38
rs536035857 Chr.3: 108560072-108560072 on Build GRCh38
rs752385051 Chr.3: 108559985-108559985 on Build GRCh38
rs202246987 Chr.3: 108559899-108559899 on Build GRCh38
rs141861595 Chr.3: 108560024-108560024 on Build GRCh38
rs751557429 Chr.3: 108560023-108560023 on Build GRCh38
rs766934851 Chr.3: 108560044-108560044 on Build GRCh38
rs768714913 Chr.3: 108560044-108560044 on Build GRCh38
rs748459686 Chr.3: 108560065-108560065 on Build GRCh38
rs766040896 Chr.3: 108559931-108559931 on Build GRCh38
rs370937449 Chr.3: 108559879-108559879 on Build GRCh38
rs376519054 Chr.3: 108559864-108559864 on Build GRCh38
rs773839482 Chr.3: 108560070-108560070 on Build GRCh38
rs965706365 Chr.3: 108559930-108559930 on Build GRCh38
rs769930293 Chr.3: 108559890-108559890 on Build GRCh38
rs762294439 Chr.3: 108559907-108559907 on Build GRCh38
rs749420545 Chr.3: 108560043-108560043 on Build GRCh38
rs780310795 Chr.3: 108559894-108559894 on Build GRCh38
rs759152178 Chr.3: 108559944-108559944 on Build GRCh38
rs575918934 Chr.3: 108560104-108560104 on Build GRCh38
rs965468339 Chr.3: 108560031-108560031 on Build GRCh38
rs772652747 Chr.3: 108559909-108559909 on Build GRCh38
rs753501920 Chr.3: 108559940-108559940 on Build GRCh38
rs948944994 Chr.3: 108560134-108560134 on Build GRCh38
rs554730788 Chr.3: 108559986-108559986 on Build GRCh38
rs200870771 Chr.3: 108559901-108559901 on Build GRCh38
rs985023241 Chr.3: 108560122-108560122 on Build GRCh38

View Full Product Details