Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 65975911-65976309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUSB
Entrez Gene ID
2990
Gene Name
glucuronidase beta
Gene Aliases
BG, MPS7
UniGene
Hs.255230
Gene Chromosome Location
Chr.7: 65960684-65982314 on Build GRCh38
Transcripts
NM_000181.3 NM_001284290.1 NM_001293104.1 NM_001293105.1 NR_120531.1 XM_005250297.3 XM_011516113.1 XM_011516114.1 XM_017012091.1 XM_017012092.1 XM_017012093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237809 c.853G>A p.V285I Chr.7: 65976074-65976074 on Build GRCh38
5191667 c.812C>T p.A271V Chr.7: 65976115-65976115 on Build GRCh38
7321116 c.882C>T p.H294H Chr.7: 65976045-65976045 on Build GRCh38
3367004 c.753G>A p.K251K Chr.7: 65976174-65976174 on Build GRCh38
3640092 c.859C>T p.L287F Chr.7: 65976068-65976068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866000884 Chr.7: 65976152-65976152 on Build GRCh38
rs753430922 Chr.7: 65976066-65976066 on Build GRCh38
rs757221229 Chr.7: 65976047-65976068 on Build GRCh38
rs764866250 Chr.7: 65976189-65976189 on Build GRCh38
rs866606409 Chr.7: 65976203-65976203 on Build GRCh38
rs777958630 Chr.7: 65976246-65976246 on Build GRCh38
rs376639818 Chr.7: 65976243-65976243 on Build GRCh38
rs894389452 Chr.7: 65975966-65975966 on Build GRCh38
rs62470936 Chr.7: 65976218-65976218 on Build GRCh38
rs183719203 Chr.7: 65975961-65975961 on Build GRCh38
rs781404289 Chr.7: 65976223-65976223 on Build GRCh38
rs751945330 Chr.7: 65976053-65976053 on Build GRCh38
rs778872193 Chr.7: 65975997-65975997 on Build GRCh38
rs377665214 Chr.7: 65976057-65976057 on Build GRCh38
rs748041603 Chr.7: 65976239-65976252 on Build GRCh38
rs569297611 Chr.7: 65976020-65976020 on Build GRCh38
rs1048020478 Chr.7: 65976021-65976021 on Build GRCh38
rs745657235 Chr.7: 65975999-65975999 on Build GRCh38
rs755827819 Chr.7: 65975990-65975990 on Build GRCh38
rs747297355 Chr.7: 65976012-65976012 on Build GRCh38
rs150086524 Chr.7: 65976167-65976167 on Build GRCh38
rs748229629 Chr.7: 65976131-65976131 on Build GRCh38
rs1057205332 Chr.7: 65976213-65976213 on Build GRCh38
rs915816962 Chr.7: 65976267-65976267 on Build GRCh38
rs140709901 Chr.7: 65976163-65976163 on Build GRCh38
rs768444459 Chr.7: 65976209-65976210 on Build GRCh38
rs763358124 Chr.7: 65976042-65976042 on Build GRCh38
rs760452541 Chr.7: 65976056-65976056 on Build GRCh38
rs762342769 Chr.7: 65975955-65975955 on Build GRCh38
rs756721679 Chr.7: 65976077-65976077 on Build GRCh38
rs774882132 Chr.7: 65976159-65976159 on Build GRCh38
rs370555813 Chr.7: 65975993-65975993 on Build GRCh38
rs748472223 Chr.7: 65976234-65976234 on Build GRCh38
rs761355885 Chr.7: 65976178-65976178 on Build GRCh38
rs769976411 Chr.7: 65976143-65976143 on Build GRCh38
rs899212988 Chr.7: 65976280-65976280 on Build GRCh38
rs765097533 Chr.7: 65976085-65976085 on Build GRCh38
rs752458446 Chr.7: 65975973-65975973 on Build GRCh38
rs371485895 Chr.7: 65976219-65976219 on Build GRCh38
rs931223727 Chr.7: 65976279-65976279 on Build GRCh38
rs758263587 Chr.7: 65976099-65976099 on Build GRCh38
rs761828939 Chr.7: 65976023-65976023 on Build GRCh38
rs771840511 Chr.7: 65976000-65976000 on Build GRCh38
rs571704452 Chr.7: 65976120-65976120 on Build GRCh38
rs773699561 Chr.7: 65976041-65976041 on Build GRCh38
rs1019311110 Chr.7: 65976095-65976095 on Build GRCh38
rs749045744 Chr.7: 65976004-65976006 on Build GRCh38
rs754699640 Chr.7: 65976220-65976220 on Build GRCh38
rs766837276 Chr.7: 65976045-65976045 on Build GRCh38
rs778741053 Chr.7: 65976230-65976233 on Build GRCh38
rs571815706 Chr.7: 65976260-65976260 on Build GRCh38
rs398123238 Chr.7: 65976061-65976061 on Build GRCh38
rs928685571 Chr.7: 65976242-65976242 on Build GRCh38
rs994841795 Chr.7: 65976281-65976281 on Build GRCh38
rs757284086 Chr.7: 65975996-65975996 on Build GRCh38
rs767091449 Chr.7: 65975972-65975972 on Build GRCh38
rs776687395 Chr.7: 65975949-65975949 on Build GRCh38
rs528899475 Chr.7: 65976040-65976040 on Build GRCh38
rs779460964 Chr.7: 65976079-65976079 on Build GRCh38
rs865932902 Chr.7: 65976110-65976110 on Build GRCh38
rs868642665 Chr.7: 65976108-65976108 on Build GRCh38
rs776736558 Chr.7: 65976022-65976022 on Build GRCh38
rs192905603 Chr.7: 65975957-65975957 on Build GRCh38
rs369465888 Chr.7: 65976115-65976115 on Build GRCh38
rs755211573 Chr.7: 65976122-65976122 on Build GRCh38
rs771320756 Chr.7: 65976148-65976148 on Build GRCh38
rs779699231 Chr.7: 65976010-65976010 on Build GRCh38
rs991304689 Chr.7: 65976245-65976245 on Build GRCh38
rs753377381 Chr.7: 65975994-65975994 on Build GRCh38
rs200941434 Chr.7: 65976161-65976161 on Build GRCh38
rs768781100 Chr.7: 65975941-65975941 on Build GRCh38
rs758405750 Chr.7: 65976216-65976216 on Build GRCh38
rs749865548 Chr.7: 65976212-65976212 on Build GRCh38
rs749282974 Chr.7: 65976250-65976250 on Build GRCh38
rs750430817 Chr.7: 65976090-65976090 on Build GRCh38
rs768075787 Chr.7: 65976146-65976146 on Build GRCh38
rs763334776 Chr.7: 65975962-65975962 on Build GRCh38
rs150999162 Chr.7: 65976114-65976114 on Build GRCh38
rs373590814 Chr.7: 65976145-65976145 on Build GRCh38

View Full Product Details