Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8206678-8206847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKB
Entrez Gene ID
9212
Gene Name
aurora kinase B
Gene Aliases
AIK2, AIM-1, AIM1, ARK2, AurB, IPL1, PPP1R48, STK12, STK5, aurkb-sv1, aurkb-sv2
UniGene
Hs.442658
Gene Chromosome Location
Chr.17: 8204731-8210768 on Build GRCh38
Transcripts
NM_001256834.2 NM_001284526.1 NM_001313950.1 NM_001313951.1 NM_001313952.1 NM_001313953.1 NM_001313954.1 NM_001313955.1 NM_004217.3 NR_132730.1 NR_132731.1 XM_011524070.2 XM_011524072.2 XM_017025307.1 XM_017025308.1 XM_017025309.1 XM_017025310.1 XM_017025311.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3691963 c.501G>A p.Q167Q Chr.17: 8206786-8206786 on Build GRCh38
5055913 c.478G>A p.G160R Chr.17: 8206809-8206809 on Build GRCh38
5924679 c.475C>T p.R159C Chr.17: 8206812-8206812 on Build GRCh38
5981142 c.529A>G p.T177A Chr.17: 8206758-8206758 on Build GRCh38
3362421 c.472C>G p.P158A Chr.17: 8206815-8206815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778473673 Chr.17: 8206803-8206803 on Build GRCh38
rs368991772 Chr.17: 8206705-8206705 on Build GRCh38
rs369322461 Chr.17: 8206731-8206731 on Build GRCh38
rs147097910 Chr.17: 8206818-8206818 on Build GRCh38
rs755736281 Chr.17: 8206724-8206724 on Build GRCh38
rs752034208 Chr.17: 8206702-8206702 on Build GRCh38
rs756423719 Chr.17: 8206768-8206768 on Build GRCh38
rs751644642 Chr.17: 8206760-8206760 on Build GRCh38
rs771962527 Chr.17: 8206742-8206742 on Build GRCh38
rs912945164 Chr.17: 8206697-8206697 on Build GRCh38
rs772022668 Chr.17: 8206809-8206809 on Build GRCh38
rs377051536 Chr.17: 8206699-8206699 on Build GRCh38
rs745673717 Chr.17: 8206811-8206812 on Build GRCh38
rs949456142 Chr.17: 8206739-8206739 on Build GRCh38
rs779807388 Chr.17: 8206810-8206810 on Build GRCh38
rs376270720 Chr.17: 8206725-8206725 on Build GRCh38
rs773526753 Chr.17: 8206746-8206746 on Build GRCh38
rs748524537 Chr.17: 8206720-8206720 on Build GRCh38
rs139322514 Chr.17: 8206761-8206761 on Build GRCh38
rs746973523 Chr.17: 8206811-8206811 on Build GRCh38
rs778145484 Chr.17: 8206775-8206775 on Build GRCh38
rs199630207 Chr.17: 8206767-8206767 on Build GRCh38
rs141770478 Chr.17: 8206788-8206788 on Build GRCh38
rs563853963 Chr.17: 8206758-8206758 on Build GRCh38
rs745437215 Chr.17: 8206808-8206808 on Build GRCh38
rs556060247 Chr.17: 8206786-8206786 on Build GRCh38
rs542566414 Chr.17: 8206741-8206741 on Build GRCh38
rs781602367 Chr.17: 8206712-8206712 on Build GRCh38
rs531140672 Chr.17: 8206812-8206812 on Build GRCh38
rs778293353 Chr.17: 8206740-8206740 on Build GRCh38
rs760595469 Chr.17: 8206745-8206745 on Build GRCh38
rs756918006 Chr.17: 8206792-8206792 on Build GRCh38
rs55871613 Chr.17: 8206751-8206751 on Build GRCh38

View Full Product Details