Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120979022-120979164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNF1A
Entrez Gene ID
6927
Gene Name
HNF1 homeobox A
Gene Aliases
HNF-1A, HNF1, IDDM20, LFB1, MODY3, TCF-1, TCF1
UniGene
Hs.654455
Gene Chromosome Location
Chr.12: 120978515-121002512 on Build GRCh38
Transcripts
NM_000545.6 NM_001306179.1 XM_005253931.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6972381 c.304G>A p.A102T Chr.12: 120979072-120979072 on Build GRCh38
46449 c.326+2_326+3delTA p.? Chr.12: 120979096-120979097 on Build GRCh38
6941177 c.305C>T p.A102V Chr.12: 120979073-120979073 on Build GRCh38
6922859 c.313G>A p.E105K Chr.12: 120979081-120979081 on Build GRCh38
4916666 c.326+2T>A p.? Chr.12: 120979096-120979096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773603125 Chr.12: 120979117-120979117 on Build GRCh38
rs199785468 Chr.12: 120979118-120979118 on Build GRCh38
rs775088578 Chr.12: 120979059-120979059 on Build GRCh38
rs80051981 Chr.12: 120979114-120979114 on Build GRCh38
rs1021001131 Chr.12: 120979143-120979143 on Build GRCh38
rs886038348 Chr.12: 120979104-120979105 on Build GRCh38
rs767920534 Chr.12: 120979119-120979119 on Build GRCh38
rs367839639 Chr.12: 120979075-120979075 on Build GRCh38
rs374794304 Chr.12: 120979058-120979058 on Build GRCh38
rs752900746 Chr.12: 120979137-120979137 on Build GRCh38
rs760168675 Chr.12: 120979111-120979111 on Build GRCh38
rs756163357 Chr.12: 120979142-120979142 on Build GRCh38
rs749659510 Chr.12: 120979102-120979103 on Build GRCh38
rs34056805 Chr.12: 120979044-120979044 on Build GRCh38
rs771474980 Chr.12: 120979136-120979137 on Build GRCh38
rs777861783 Chr.12: 120979145-120979145 on Build GRCh38
rs966714279 Chr.12: 120979053-120979053 on Build GRCh38
rs892943642 Chr.12: 120979140-120979141 on Build GRCh38
rs937297373 Chr.12: 120979120-120979120 on Build GRCh38
rs775109216 Chr.12: 120979108-120979108 on Build GRCh38
rs774971768 Chr.12: 120979141-120979141 on Build GRCh38
rs56158114 Chr.12: 120979099-120979100 on Build GRCh38
rs941731027 Chr.12: 120979079-120979080 on Build GRCh38
rs193922593 Chr.12: 120979049-120979049 on Build GRCh38
rs370313011 Chr.12: 120979105-120979105 on Build GRCh38
rs774093881 Chr.12: 120979066-120979066 on Build GRCh38
rs193922594 Chr.12: 120979081-120979082 on Build GRCh38
rs1800574 Chr.12: 120979061-120979061 on Build GRCh38
rs193922595 Chr.12: 120979098-120979098 on Build GRCh38

View Full Product Details