Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5050752-5050922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSL6
Entrez Gene ID
11172
Gene Name
insulin like 6
Gene Aliases
RIF1
UniGene
Hs.632648
Gene Chromosome Location
Chr.9: 4992408-5185706 on Build GRCh38
Transcripts
XM_011517702.2 NM_001322194.1 NM_001322195.1 NM_001322196.1 NM_001322198.1 NM_001322199.1 NM_001322204.1 NM_004972.3

Gene Symbol
JAK2
Entrez Gene ID
3717
Gene Name
Janus kinase 2
Gene Aliases
JTK10, THCYT3
UniGene
Hs.656213
Gene Chromosome Location
Chr.9: 4985086-5128183 on Build GRCh38
Transcripts
XM_011517702.2 NM_001322194.1 NM_001322195.1 NM_001322196.1 NM_001322198.1 NM_001322199.1 NM_001322204.1 NM_004972.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5095532 c.548T>G p.V183G Chr.9: 5050765-5050765 on Build GRCh38
6912177 c.604_612delAACTCTATC p.N202_I204delNSI Chr.9: 5050821-5050829 on Build GRCh38
6939271 c.544G>A p.A182T Chr.9: 5050761-5050761 on Build GRCh38
4840834 c.574G>T p.E192* Chr.9: 5050791-5050791 on Build GRCh38
5609189 c.608C>T p.S203F Chr.9: 5050825-5050825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540747820 Chr.9: 5050798-5050798 on Build GRCh38
rs532894314 Chr.9: 5050819-5050819 on Build GRCh38
rs916171915 Chr.9: 5050894-5050894 on Build GRCh38
rs192178119 Chr.9: 5050864-5050864 on Build GRCh38
rs989983120 Chr.9: 5050858-5050858 on Build GRCh38
rs761496056 Chr.9: 5050851-5050851 on Build GRCh38
rs938904337 Chr.9: 5050889-5050889 on Build GRCh38
rs770883479 Chr.9: 5050778-5050778 on Build GRCh38
rs752012407 Chr.9: 5050836-5050836 on Build GRCh38
rs142724240 Chr.9: 5050879-5050879 on Build GRCh38
rs375180351 Chr.9: 5050827-5050827 on Build GRCh38
rs754815036 Chr.9: 5050868-5050868 on Build GRCh38
rs745954795 Chr.9: 5050789-5050789 on Build GRCh38
rs41302214 Chr.9: 5050796-5050796 on Build GRCh38
rs749901402 Chr.9: 5050857-5050857 on Build GRCh38
rs559371331 Chr.9: 5050809-5050809 on Build GRCh38
rs753837462 Chr.9: 5050840-5050840 on Build GRCh38
rs1056036845 Chr.9: 5050890-5050890 on Build GRCh38
rs376697477 Chr.9: 5050816-5050816 on Build GRCh38
rs928672000 Chr.9: 5050888-5050888 on Build GRCh38
rs781379515 Chr.9: 5050782-5050782 on Build GRCh38
rs187871601 Chr.9: 5050856-5050856 on Build GRCh38
rs935518281 Chr.9: 5050803-5050803 on Build GRCh38
rs963896092 Chr.9: 5050846-5050846 on Build GRCh38
rs766273126 Chr.9: 5050863-5050863 on Build GRCh38
rs369119377 Chr.9: 5050859-5050859 on Build GRCh38

View Full Product Details