Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 95107017-95107172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf3
Entrez Gene ID
84909
Gene Name
chromosome 9 open reading frame 3
Gene Aliases
AOPEP, AP-O, APO, C90RF3, ONPEP
UniGene
Hs.434253
Gene Chromosome Location
Chr.9: 94726604-95148264 on Build GRCh38
Transcripts
XM_011519121.2 NM_000136.2 NM_001243743.1 XM_005251802.3 XM_006717001.2 XM_011518365.2 XM_011518367.2 XM_017014452.1 XM_017014453.1 XM_017014454.1

Gene Symbol
FANCC
Entrez Gene ID
2176
Gene Name
Fanconi anemia complementation group C
Gene Aliases
FA3, FAC, FACC
UniGene
Hs.494529
Gene Chromosome Location
Chr.9: 95099054-95317730 on Build GRCh38
Transcripts
XM_011519121.2 NM_000136.2 NM_001243743.1 XM_005251802.3 XM_006717001.2 XM_011518365.2 XM_011518367.2 XM_017014452.1 XM_017014453.1 XM_017014454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6193931 c.1511T>C p.I504T Chr.9: 95107088-95107088 on Build GRCh38
4609475 c.1483C>A p.L495M Chr.9: 95107116-95107116 on Build GRCh38
3849147 c.1466A>C p.H489P Chr.9: 95107133-95107133 on Build GRCh38
6986072 c.1456C>G p.L486V Chr.9: 95107143-95107143 on Build GRCh38
5064176 c.1513G>A p.A505T Chr.9: 95107086-95107086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576831650 Chr.9: 95107108-95107108 on Build GRCh38
rs1057517170 Chr.9: 95107064-95107064 on Build GRCh38
rs761050718 Chr.9: 95107143-95107143 on Build GRCh38
rs1057516963 Chr.9: 95107101-95107101 on Build GRCh38
rs200423281 Chr.9: 95107117-95107117 on Build GRCh38
rs762501862 Chr.9: 95107109-95107109 on Build GRCh38
rs900179723 Chr.9: 95107099-95107099 on Build GRCh38
rs746940880 Chr.9: 95107087-95107087 on Build GRCh38
rs767852720 Chr.9: 95107050-95107050 on Build GRCh38
rs863224323 Chr.9: 95107057-95107057 on Build GRCh38
rs56082100 Chr.9: 95107114-95107114 on Build GRCh38
rs730881725 Chr.9: 95107106-95107106 on Build GRCh38
rs767996832 Chr.9: 95107128-95107128 on Build GRCh38
rs1024371711 Chr.9: 95107094-95107094 on Build GRCh38
rs372199352 Chr.9: 95107069-95107069 on Build GRCh38
rs765766432 Chr.9: 95107075-95107075 on Build GRCh38
rs746540247 Chr.9: 95107098-95107098 on Build GRCh38
rs760046823 Chr.9: 95107120-95107120 on Build GRCh38
rs753885687 Chr.9: 95107065-95107065 on Build GRCh38
rs754079401 Chr.9: 95107153-95107153 on Build GRCh38
rs730881726 Chr.9: 95107083-95107083 on Build GRCh38
rs759936400 Chr.9: 95107049-95107049 on Build GRCh38
rs764271678 Chr.9: 95107149-95107149 on Build GRCh38
rs779982610 Chr.9: 95107091-95107091 on Build GRCh38
rs536599109 Chr.9: 95107097-95107097 on Build GRCh38
rs121917785 Chr.9: 95107112-95107112 on Build GRCh38
rs76895298 Chr.9: 95107105-95107105 on Build GRCh38
rs780179187 Chr.9: 95107086-95107086 on Build GRCh38
rs1057516488 Chr.9: 95107082-95107082 on Build GRCh38
rs753031127 Chr.9: 95107142-95107142 on Build GRCh38
rs144278080 Chr.9: 95107090-95107090 on Build GRCh38
rs773270231 Chr.9: 95107153-95107155 on Build GRCh38
rs200515307 Chr.9: 95107053-95107053 on Build GRCh38
rs587779900 Chr.9: 95107110-95107110 on Build GRCh38
rs756290316 Chr.9: 95107051-95107051 on Build GRCh38

View Full Product Details