Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2917202-2917329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD11
Entrez Gene ID
84433
Gene Name
caspase recruitment domain family member 11
Gene Aliases
BENTA, BIMP3, CARMA1, IMD11, PPBL
UniGene
Hs.648101
Gene Chromosome Location
Chr.7: 2906075-3043945 on Build GRCh38
Transcripts
NM_001324281.1 NM_032415.5 XM_011515586.2 XM_011515587.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5710537 c.2677A>T p.S893C Chr.7: 2917316-2917316 on Build GRCh38
6912913 c.2698C>T p.L900F Chr.7: 2917295-2917295 on Build GRCh38
5019356 c.2703+6G>T p.? Chr.7: 2917284-2917284 on Build GRCh38
5608005 c.2682C>T p.F894F Chr.7: 2917311-2917311 on Build GRCh38
1199565 c.2672G>A p.R891Q Chr.7: 2917321-2917321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377702889 Chr.7: 2917238-2917238 on Build GRCh38
rs558286041 Chr.7: 2917277-2917277 on Build GRCh38
rs753142060 Chr.7: 2917273-2917273 on Build GRCh38
rs201282256 Chr.7: 2917267-2917267 on Build GRCh38
rs539279952 Chr.7: 2917265-2917265 on Build GRCh38
rs886506846 Chr.7: 2917296-2917296 on Build GRCh38
rs191837050 Chr.7: 2917289-2917289 on Build GRCh38
rs1009103943 Chr.7: 2917231-2917231 on Build GRCh38
rs775406780 Chr.7: 2917237-2917237 on Build GRCh38
rs772771059 Chr.7: 2917253-2917253 on Build GRCh38
rs3735123 Chr.7: 2917284-2917284 on Build GRCh38
rs370020664 Chr.7: 2917243-2917243 on Build GRCh38
rs375163437 Chr.7: 2917266-2917266 on Build GRCh38
rs201256077 Chr.7: 2917286-2917286 on Build GRCh38
rs556715404 Chr.7: 2917239-2917239 on Build GRCh38
rs372718712 Chr.7: 2917278-2917278 on Build GRCh38
rs3735122 Chr.7: 2917250-2917250 on Build GRCh38
rs955849261 Chr.7: 2917232-2917232 on Build GRCh38
rs772298133 Chr.7: 2917284-2917284 on Build GRCh38
rs902761185 Chr.7: 2917229-2917229 on Build GRCh38
rs369243746 Chr.7: 2917271-2917271 on Build GRCh38
rs781316312 Chr.7: 2917299-2917299 on Build GRCh38

View Full Product Details