Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127261174-127261298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC3
Entrez Gene ID
2071
Gene Name
ERCC excision repair 3, TFIIH core complex helicase subunit
Gene Aliases
BTF2, GTF2H, RAD25, TFIIH, TTD2, XPB
UniGene
Hs.469872
Gene Chromosome Location
Chr.2: 127257290-127294176 on Build GRCh38
Transcripts
NM_000122.1 NM_001303416.1 NM_001303418.1 XM_011510794.1 XM_011510795.1 XM_017003583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6925754 c.2034A>T p.R678S Chr.2: 127261258-127261258 on Build GRCh38
6912822 c.2011G>T p.A671S Chr.2: 127261281-127261281 on Build GRCh38
5707754 c.2032A>G p.R678G Chr.2: 127261260-127261260 on Build GRCh38
5860055 c.1994A>T p.Q665L Chr.2: 127261298-127261298 on Build GRCh38
5965738 c.2013T>C p.A671A Chr.2: 127261279-127261279 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41553418 Chr.2: 127261264-127261264 on Build GRCh38
rs775187800 Chr.2: 127261239-127261239 on Build GRCh38
rs746280448 Chr.2: 127261241-127261241 on Build GRCh38
rs372749473 Chr.2: 127261223-127261223 on Build GRCh38
rs760140217 Chr.2: 127261258-127261259 on Build GRCh38
rs768284835 Chr.2: 127261272-127261272 on Build GRCh38
rs922942314 Chr.2: 127261266-127261266 on Build GRCh38
rs771388157 Chr.2: 127261196-127261196 on Build GRCh38
rs375990467 Chr.2: 127261274-127261274 on Build GRCh38
rs975748269 Chr.2: 127261224-127261224 on Build GRCh38
rs375844247 Chr.2: 127261225-127261225 on Build GRCh38
rs1023234786 Chr.2: 127261205-127261205 on Build GRCh38
rs368196350 Chr.2: 127261195-127261195 on Build GRCh38
rs767218456 Chr.2: 127261208-127261209 on Build GRCh38

View Full Product Details