Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57749162-57749327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK4
Entrez Gene ID
1019
Gene Name
cyclin dependent kinase 4
Gene Aliases
CMM3, PSK-J3
UniGene
Hs.95577
Gene Chromosome Location
Chr.12: 57747727-57752447 on Build GRCh38
Transcripts
NM_000075.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1989827 c.740G>A p.G247E Chr.12: 57749261-57749261 on Build GRCh38
6929125 c.721G>A p.D241N Chr.12: 57749280-57749280 on Build GRCh38
1163016 c.736C>T p.R246C Chr.12: 57749265-57749265 on Build GRCh38
6937557 c.784C>T p.P262S Chr.12: 57749217-57749217 on Build GRCh38
4507017 c.735C>T p.P245P Chr.12: 57749266-57749266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755502728 Chr.12: 57749280-57749280 on Build GRCh38
rs144657355 Chr.12: 57749237-57749237 on Build GRCh38
rs786203856 Chr.12: 57749283-57749283 on Build GRCh38
rs786202902 Chr.12: 57749196-57749196 on Build GRCh38
rs748377822 Chr.12: 57749218-57749218 on Build GRCh38
rs368081942 Chr.12: 57749299-57749299 on Build GRCh38
rs377612647 Chr.12: 57749230-57749230 on Build GRCh38
rs876660217 Chr.12: 57749273-57749273 on Build GRCh38
rs374223296 Chr.12: 57749296-57749296 on Build GRCh38
rs531817742 Chr.12: 57749282-57749282 on Build GRCh38
rs201617914 Chr.12: 57749225-57749225 on Build GRCh38
rs747362286 Chr.12: 57749276-57749276 on Build GRCh38
rs201744062 Chr.12: 57749200-57749200 on Build GRCh38
rs876658903 Chr.12: 57749277-57749277 on Build GRCh38
rs3211622 Chr.12: 57749224-57749224 on Build GRCh38
rs2227953 Chr.12: 57749305-57749305 on Build GRCh38
rs921450127 Chr.12: 57749261-57749261 on Build GRCh38
rs772654769 Chr.12: 57749215-57749215 on Build GRCh38
rs762519743 Chr.12: 57749216-57749216 on Build GRCh38
rs200215596 Chr.12: 57749222-57749222 on Build GRCh38
rs752435107 Chr.12: 57749233-57749233 on Build GRCh38
rs779161525 Chr.12: 57749195-57749195 on Build GRCh38
rs587778188 Chr.12: 57749238-57749238 on Build GRCh38
rs780522588 Chr.12: 57749203-57749203 on Build GRCh38
rs757333818 Chr.12: 57749248-57749248 on Build GRCh38
rs769226197 Chr.12: 57749208-57749208 on Build GRCh38
rs876660340 Chr.12: 57749223-57749223 on Build GRCh38
rs886049713 Chr.12: 57749254-57749254 on Build GRCh38
rs753908111 Chr.12: 57749239-57749239 on Build GRCh38
rs770507806 Chr.12: 57749295-57749295 on Build GRCh38
rs754187815 Chr.12: 57749247-57749248 on Build GRCh38
rs771702415 Chr.12: 57749300-57749300 on Build GRCh38
rs143670820 Chr.12: 57749249-57749249 on Build GRCh38
rs587779896 Chr.12: 57749241-57749241 on Build GRCh38
rs370258992 Chr.12: 57749265-57749265 on Build GRCh38
rs372604524 Chr.12: 57749264-57749264 on Build GRCh38
rs747521511 Chr.12: 57749206-57749206 on Build GRCh38

View Full Product Details