Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127257500-127257636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC3
Entrez Gene ID
2071
Gene Name
ERCC excision repair 3, TFIIH core complex helicase subunit
Gene Aliases
BTF2, GTF2H, RAD25, TFIIH, TTD2, XPB
UniGene
Hs.469872
Gene Chromosome Location
Chr.2: 127257290-127294176 on Build GRCh38
Transcripts
NM_000122.1 NM_001303416.1 NM_001303418.1 XM_011510794.1 XM_011510795.1 XM_017003583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5954310 c.2335C>T p.R779C Chr.2: 127257610-127257610 on Build GRCh38
6920857 c.2331C>G p.F777L Chr.2: 127257614-127257614 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758104976 Chr.2: 127257591-127257591 on Build GRCh38
rs146427260 Chr.2: 127257609-127257609 on Build GRCh38
rs200411611 Chr.2: 127257604-127257604 on Build GRCh38
rs745534678 Chr.2: 127257584-127257584 on Build GRCh38
rs766339532 Chr.2: 127257547-127257547 on Build GRCh38
rs763049905 Chr.2: 127257546-127257546 on Build GRCh38
rs202107588 Chr.2: 127257577-127257577 on Build GRCh38
rs753046922 Chr.2: 127257570-127257570 on Build GRCh38
rs4150523 Chr.2: 127257567-127257567 on Build GRCh38
rs751739122 Chr.2: 127257564-127257564 on Build GRCh38
rs755532523 Chr.2: 127257566-127257566 on Build GRCh38
rs147499527 Chr.2: 127257611-127257611 on Build GRCh38
rs766592763 Chr.2: 127257610-127257610 on Build GRCh38

View Full Product Details