Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 28695069-28695234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHEK2
Entrez Gene ID
11200
Gene Name
checkpoint kinase 2
Gene Aliases
CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1
UniGene
Hs.291363
Gene Chromosome Location
Chr.22: 28687743-28741866 on Build GRCh38
Transcripts
NM_001005735.1 NM_001257387.1 NM_007194.3 NM_145862.2 XM_006724114.3 XM_006724116.2 XM_011529839.2 XM_011529840.2 XM_011529841.1 XM_011529842.2 XM_011529843.1 XM_011529845.2 XM_017028560.1 XM_017028561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361160 c.1343T>G p.I448S Chr.22: 28695159-28695159 on Build GRCh38
6980423 c.1406C>T p.P469L Chr.22: 28695225-28695225 on Build GRCh38
7361159 c.1472T>G p.I491S Chr.22: 28695159-28695159 on Build GRCh38
4771012 c.1367C>G p.S456* Chr.22: 28695135-28695135 on Build GRCh38
6947115 c.1498G>A p.E500K Chr.22: 28695133-28695133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750012749 Chr.22: 28695160-28695160 on Build GRCh38
rs759706360 Chr.22: 28695126-28695126 on Build GRCh38
rs876659639 Chr.22: 28695165-28695165 on Build GRCh38
rs878854914 Chr.22: 28695195-28695195 on Build GRCh38
rs876660046 Chr.22: 28695181-28695181 on Build GRCh38
rs763395924 Chr.22: 28695145-28695145 on Build GRCh38
rs776700371 Chr.22: 28695111-28695111 on Build GRCh38
rs527418829 Chr.22: 28695101-28695101 on Build GRCh38
rs876660801 Chr.22: 28695125-28695126 on Build GRCh38
rs778246892 Chr.22: 28695168-28695168 on Build GRCh38
rs876659868 Chr.22: 28695124-28695124 on Build GRCh38
rs786201687 Chr.22: 28695158-28695158 on Build GRCh38
rs121908705 Chr.22: 28695166-28695166 on Build GRCh38
rs730881702 Chr.22: 28695146-28695146 on Build GRCh38
rs747239326 Chr.22: 28695096-28695096 on Build GRCh38
rs139088611 Chr.22: 28695150-28695150 on Build GRCh38
rs17886163 Chr.22: 28695159-28695159 on Build GRCh38
rs548796284 Chr.22: 28695151-28695151 on Build GRCh38
rs876659827 Chr.22: 28695135-28695135 on Build GRCh38
rs753839004 Chr.22: 28695179-28695179 on Build GRCh38
rs730881700 Chr.22: 28695133-28695134 on Build GRCh38
rs778791004 Chr.22: 28695094-28695094 on Build GRCh38
rs17882922 Chr.22: 28695196-28695196 on Build GRCh38
rs878854915 Chr.22: 28695185-28695185 on Build GRCh38
rs779383555 Chr.22: 28695177-28695177 on Build GRCh38
rs17880867 Chr.22: 28695164-28695164 on Build GRCh38
rs771772104 Chr.22: 28695211-28695212 on Build GRCh38
rs776093527 Chr.22: 28695128-28695128 on Build GRCh38
rs876660323 Chr.22: 28695202-28695202 on Build GRCh38
rs200050883 Chr.22: 28695190-28695190 on Build GRCh38
rs17881473 Chr.22: 28695163-28695163 on Build GRCh38
rs769729382 Chr.22: 28695127-28695127 on Build GRCh38
rs764449869 Chr.22: 28695148-28695148 on Build GRCh38
rs778989252 Chr.22: 28695187-28695187 on Build GRCh38
rs587780175 Chr.22: 28695209-28695209 on Build GRCh38
rs587778194 Chr.22: 28695169-28695169 on Build GRCh38
rs587780176 Chr.22: 28695184-28695184 on Build GRCh38
rs373864492 Chr.22: 28695212-28695212 on Build GRCh38
rs876660373 Chr.22: 28695162-28695162 on Build GRCh38

View Full Product Details