Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44107879-44108031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXT2
Entrez Gene ID
2132
Gene Name
exostosin glycosyltransferase 2
Gene Aliases
SOTV, SSMS
UniGene
Hs.368404
Gene Chromosome Location
Chr.11: 44095549-44245430 on Build GRCh38
Transcripts
NM_000401.3 NM_001178083.1 NM_207122.1 XM_011519950.1 XM_011519951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
94262 c.257G>C p.R86T Chr.11: 44107969-44107969 on Build GRCh38
7295253 c.238C>T p.R80W Chr.11: 44107950-44107950 on Build GRCh38
1243234 c.266C>T p.T89M Chr.11: 44107978-44107978 on Build GRCh38
224982 c.283C>T p.R95C Chr.11: 44107995-44107995 on Build GRCh38
5891437 c.284G>A p.R95H Chr.11: 44107996-44107996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746889702 Chr.11: 44107977-44107977 on Build GRCh38
rs112231770 Chr.11: 44107998-44107998 on Build GRCh38
rs143048174 Chr.11: 44107978-44107978 on Build GRCh38
rs759924417 Chr.11: 44107932-44107932 on Build GRCh38
rs745846072 Chr.11: 44107902-44107902 on Build GRCh38
rs143703574 Chr.11: 44107996-44107996 on Build GRCh38
rs140075817 Chr.11: 44107972-44107972 on Build GRCh38
rs200809171 Chr.11: 44107917-44107917 on Build GRCh38
rs758194845 Chr.11: 44107973-44107973 on Build GRCh38
rs534539796 Chr.11: 44107957-44107957 on Build GRCh38
rs35455466 Chr.11: 44107976-44107976 on Build GRCh38
rs376292686 Chr.11: 44107995-44107995 on Build GRCh38
rs555206160 Chr.11: 44107903-44107903 on Build GRCh38
rs763536229 Chr.11: 44107951-44107951 on Build GRCh38
rs774620075 Chr.11: 44107931-44107931 on Build GRCh38
rs999897934 Chr.11: 44107938-44107938 on Build GRCh38
rs768071412 Chr.11: 44107939-44107939 on Build GRCh38
rs755625888 Chr.11: 44107950-44107950 on Build GRCh38
rs773387959 Chr.11: 44107990-44107990 on Build GRCh38
rs868482302 Chr.11: 44107943-44107943 on Build GRCh38
rs587778299 Chr.11: 44107944-44107944 on Build GRCh38
rs559280035 Chr.11: 44107958-44107958 on Build GRCh38
rs763040366 Chr.11: 44107922-44107922 on Build GRCh38
rs773585874 Chr.11: 44107920-44107920 on Build GRCh38
rs920725366 Chr.11: 44107999-44107999 on Build GRCh38
rs766800368 Chr.11: 44107923-44107923 on Build GRCh38
rs201374853 Chr.11: 44107913-44107913 on Build GRCh38
rs748292026 Chr.11: 44107912-44107912 on Build GRCh38
rs778594219 Chr.11: 44107965-44107965 on Build GRCh38
rs775542605 Chr.11: 44107910-44107910 on Build GRCh38

View Full Product Details