Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56370092-56370232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKA
Entrez Gene ID
6790
Gene Name
aurora kinase A
Gene Aliases
AIK, ARK1, AURA, BTAK, PPP1R47, STK15, STK6, STK7
UniGene
Hs.250822
Gene Chromosome Location
Chr.20: 56369389-56392337 on Build GRCh38
Transcripts
NM_001323303.1 NM_001323304.1 NM_001323305.1 NM_003600.3 NM_198433.2 NM_198434.2 NM_198435.2 NM_198436.2 NM_198437.2 XM_017028034.1 XM_017028035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6958988 c.1163C>G p.S388* Chr.20: 56370207-56370207 on Build GRCh38
2763898 c.1149C>A p.I383I Chr.20: 56370221-56370221 on Build GRCh38
6954237 c.1186A>G p.K396E Chr.20: 56370184-56370184 on Build GRCh38
5569879 c.1152A>G p.T384T Chr.20: 56370218-56370218 on Build GRCh38
6950986 c.1151C>G p.T384R Chr.20: 56370219-56370219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113451501 Chr.20: 56370186-56370186 on Build GRCh38
rs778256805 Chr.20: 56370165-56370165 on Build GRCh38
rs34361943 Chr.20: 56370155-56370155 on Build GRCh38
rs1051125021 Chr.20: 56370149-56370149 on Build GRCh38
rs764540655 Chr.20: 56370201-56370201 on Build GRCh38
rs745433550 Chr.20: 56370166-56370166 on Build GRCh38
rs201506695 Chr.20: 56370143-56370143 on Build GRCh38
rs777727345 Chr.20: 56370152-56370152 on Build GRCh38
rs754845385 Chr.20: 56370120-56370120 on Build GRCh38
rs536637669 Chr.20: 56370211-56370211 on Build GRCh38
rs376893456 Chr.20: 56370199-56370199 on Build GRCh38
rs762455284 Chr.20: 56370200-56370201 on Build GRCh38
rs752715782 Chr.20: 56370142-56370142 on Build GRCh38
rs761905670 Chr.20: 56370196-56370196 on Build GRCh38
rs775906971 Chr.20: 56370177-56370177 on Build GRCh38
rs747528451 Chr.20: 56370178-56370178 on Build GRCh38
rs771512723 Chr.20: 56370171-56370171 on Build GRCh38
rs781138597 Chr.20: 56370124-56370124 on Build GRCh38
rs755910157 Chr.20: 56370146-56370146 on Build GRCh38
rs374112531 Chr.20: 56370133-56370133 on Build GRCh38
rs35436934 Chr.20: 56370153-56370153 on Build GRCh38
rs56018429 Chr.20: 56370190-56370190 on Build GRCh38
rs1037323685 Chr.20: 56370164-56370164 on Build GRCh38
rs573740861 Chr.20: 56370191-56370191 on Build GRCh38

View Full Product Details