Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3526059-3526202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZR1
Entrez Gene ID
51343
Gene Name
fizzy/cell division cycle 20 related 1
Gene Aliases
CDC20C, CDH1, FZR, FZR2, HCDH, HCDH1
UniGene
Hs.413133
Gene Chromosome Location
Chr.19: 3506115-3536757 on Build GRCh38
Transcripts
NM_001136197.1 NM_001136198.1 NM_016263.3 XM_005259573.4 XM_017026863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4281469 c.209C>T p.S70F Chr.19: 3526133-3526133 on Build GRCh38
5674841 c.211C>G p.P71A Chr.19: 3526135-3526135 on Build GRCh38
3197028 c.209C>T p.S70F Chr.19: 3526133-3526133 on Build GRCh38
6897680 c.219G>A p.Q73Q Chr.19: 3526143-3526143 on Build GRCh38
3532355 c.252C>A p.N84K Chr.19: 3526176-3526176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754355403 Chr.19: 3526177-3526177 on Build GRCh38
rs769401817 Chr.19: 3526111-3526111 on Build GRCh38
rs146055485 Chr.19: 3526162-3526162 on Build GRCh38
rs575076320 Chr.19: 3526091-3526091 on Build GRCh38
rs142885173 Chr.19: 3526161-3526161 on Build GRCh38
rs754337303 Chr.19: 3526087-3526087 on Build GRCh38
rs759834366 Chr.19: 3526148-3526148 on Build GRCh38
rs900829580 Chr.19: 3526155-3526155 on Build GRCh38
rs139197779 Chr.19: 3526130-3526130 on Build GRCh38
rs201608894 Chr.19: 3526107-3526107 on Build GRCh38
rs992674016 Chr.19: 3526109-3526109 on Build GRCh38
rs757696153 Chr.19: 3526088-3526088 on Build GRCh38
rs747713310 Chr.19: 3526110-3526110 on Build GRCh38
rs757634430 Chr.19: 3526179-3526179 on Build GRCh38
rs374041533 Chr.19: 3526176-3526176 on Build GRCh38
rs375691374 Chr.19: 3526080-3526080 on Build GRCh38
rs773353105 Chr.19: 3526114-3526114 on Build GRCh38
rs374299885 Chr.19: 3526173-3526173 on Build GRCh38
rs778291066 Chr.19: 3526083-3526083 on Build GRCh38
rs754542868 Chr.19: 3526104-3526104 on Build GRCh38
rs945306132 Chr.19: 3526105-3526105 on Build GRCh38
rs368628520 Chr.19: 3526089-3526089 on Build GRCh38
rs749172332 Chr.19: 3526122-3526122 on Build GRCh38
rs752733834 Chr.19: 3526077-3526077 on Build GRCh38

View Full Product Details