Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 61500318-61500486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH20
Entrez Gene ID
28316
Gene Name
cadherin 20
Gene Aliases
CDH7L3, Cdh7
UniGene
Hs.671510
Gene Chromosome Location
Chr.18: 61333582-61555773 on Build GRCh38
Transcripts
NM_031891.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4131043 c.584C>T p.P195L Chr.18: 61500425-61500425 on Build GRCh38
5697500 c.637T>C p.Y213H Chr.18: 61500478-61500478 on Build GRCh38
359221 c.573T>A p.D191E Chr.18: 61500414-61500414 on Build GRCh38
5722763 c.625C>T p.Q209* Chr.18: 61500466-61500466 on Build GRCh38
1200450 c.592G>A p.G198S Chr.18: 61500433-61500433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747619915 Chr.18: 61500417-61500417 on Build GRCh38
rs757072442 Chr.18: 61500367-61500367 on Build GRCh38
rs763468145 Chr.18: 61500356-61500356 on Build GRCh38
rs781105714 Chr.18: 61500373-61500373 on Build GRCh38
rs775307745 Chr.18: 61500452-61500452 on Build GRCh38
rs373056408 Chr.18: 61500432-61500432 on Build GRCh38
rs1049995098 Chr.18: 61500343-61500343 on Build GRCh38
rs773099480 Chr.18: 61500416-61500416 on Build GRCh38
rs867559385 Chr.18: 61500459-61500459 on Build GRCh38
rs199623629 Chr.18: 61500391-61500391 on Build GRCh38
rs781593697 Chr.18: 61500446-61500446 on Build GRCh38
rs752298835 Chr.18: 61500342-61500342 on Build GRCh38
rs772160601 Chr.18: 61500415-61500415 on Build GRCh38
rs145920224 Chr.18: 61500390-61500390 on Build GRCh38
rs542534614 Chr.18: 61500463-61500463 on Build GRCh38
rs867387320 Chr.18: 61500365-61500365 on Build GRCh38
rs762641363 Chr.18: 61500453-61500453 on Build GRCh38
rs761866120 Chr.18: 61500460-61500460 on Build GRCh38
rs937950352 Chr.18: 61500398-61500398 on Build GRCh38
rs757504605 Chr.18: 61500432-61500434 on Build GRCh38
rs894967492 Chr.18: 61500437-61500437 on Build GRCh38
rs376416616 Chr.18: 61500380-61500380 on Build GRCh38
rs200877732 Chr.18: 61500457-61500457 on Build GRCh38
rs770246700 Chr.18: 61500435-61500435 on Build GRCh38
rs941053881 Chr.18: 61500371-61500371 on Build GRCh38
rs771330895 Chr.18: 61500425-61500425 on Build GRCh38
rs748328095 Chr.18: 61500404-61500404 on Build GRCh38
rs142200700 Chr.18: 61500348-61500348 on Build GRCh38
rs150175200 Chr.18: 61500426-61500426 on Build GRCh38
rs373206188 Chr.18: 61500358-61500358 on Build GRCh38

View Full Product Details