Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8204971-8205119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKB
Entrez Gene ID
9212
Gene Name
aurora kinase B
Gene Aliases
AIK2, AIM-1, AIM1, ARK2, AurB, IPL1, PPP1R48, STK12, STK5, aurkb-sv1, aurkb-sv2
UniGene
Hs.442658
Gene Chromosome Location
Chr.17: 8204731-8210768 on Build GRCh38
Transcripts
NM_001256834.2 NM_001284526.1 NM_001313950.1 NM_001313951.1 NM_001313952.1 NM_001313953.1 NM_001313954.1 NM_001313955.1 NM_004217.3 NR_132730.1 NR_132731.1 XM_011524070.2 XM_011524072.2 XM_017025307.1 XM_017025308.1 XM_017025309.1 XM_017025310.1 XM_017025311.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6608866 c.912C>T p.I304I Chr.17: 8204994-8204994 on Build GRCh38
4664754 c.895G>A p.G299R Chr.17: 8205011-8205011 on Build GRCh38
986221 c.880G>A p.A294T Chr.17: 8205026-8205026 on Build GRCh38
6973711 c.862-2A>G p.? Chr.17: 8205046-8205046 on Build GRCh38
1387617 c.894G>T p.M298I Chr.17: 8205012-8205012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987905457 Chr.17: 8205003-8205003 on Build GRCh38
rs767082017 Chr.17: 8205029-8205029 on Build GRCh38
rs191375912 Chr.17: 8205073-8205073 on Build GRCh38
rs760974257 Chr.17: 8205005-8205005 on Build GRCh38
rs939607713 Chr.17: 8205075-8205075 on Build GRCh38
rs775624172 Chr.17: 8205001-8205001 on Build GRCh38
rs768924008 Chr.17: 8205084-8205084 on Build GRCh38
rs993055729 Chr.17: 8205048-8205048 on Build GRCh38
rs756954056 Chr.17: 8205072-8205072 on Build GRCh38
rs772308526 Chr.17: 8204999-8204999 on Build GRCh38
rs373131785 Chr.17: 8205069-8205069 on Build GRCh38
rs756119378 Chr.17: 8205051-8205051 on Build GRCh38
rs1059476 Chr.17: 8205013-8205013 on Build GRCh38
rs773593110 Chr.17: 8205025-8205025 on Build GRCh38
rs199773518 Chr.17: 8205078-8205078 on Build GRCh38
rs146334050 Chr.17: 8204995-8204995 on Build GRCh38
rs749106198 Chr.17: 8204998-8204998 on Build GRCh38
rs777899034 Chr.17: 8205064-8205064 on Build GRCh38
rs2241909 Chr.17: 8205021-8205021 on Build GRCh38
rs754686281 Chr.17: 8205027-8205027 on Build GRCh38
rs752317012 Chr.17: 8205036-8205036 on Build GRCh38
rs371287619 Chr.17: 8205070-8205070 on Build GRCh38
rs3200163 Chr.17: 8204994-8204994 on Build GRCh38
rs768656332 Chr.17: 8205012-8205012 on Build GRCh38
rs776687153 Chr.17: 8205093-8205093 on Build GRCh38
rs151173438 Chr.17: 8205026-8205026 on Build GRCh38

View Full Product Details