Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190366800-190367055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP1
Entrez Gene ID
3628
Gene Name
inositol polyphosphate-1-phosphatase
Gene Aliases
-
UniGene
Hs.32309
Gene Chromosome Location
Chr.2: 190343470-190371665 on Build GRCh38
Transcripts
NM_001128928.1 NM_002194.3 XM_005246532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4902475 c.413C>T p.S138F Chr.2: 190366842-190366842 on Build GRCh38
5956129 c.418G>A p.E140K Chr.2: 190366847-190366847 on Build GRCh38
3709279 c.425A>G p.N142S Chr.2: 190366854-190366854 on Build GRCh38
4692540 c.466+2T>C p.? Chr.2: 190366897-190366897 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760644724 Chr.2: 190366895-190366895 on Build GRCh38
rs781521573 Chr.2: 190366942-190366942 on Build GRCh38
rs747397425 Chr.2: 190366931-190366931 on Build GRCh38
rs779557122 Chr.2: 190366872-190366873 on Build GRCh38
rs748727151 Chr.2: 190366853-190366853 on Build GRCh38
rs932141799 Chr.2: 190366861-190366861 on Build GRCh38
rs1016143631 Chr.2: 190366940-190366940 on Build GRCh38
rs773141973 Chr.2: 190366893-190366893 on Build GRCh38
rs1055984350 Chr.2: 190366868-190366868 on Build GRCh38
rs747456953 Chr.2: 190366834-190366834 on Build GRCh38
rs747848074 Chr.2: 190366942-190366942 on Build GRCh38
rs757691731 Chr.2: 190366934-190366934 on Build GRCh38
rs896014246 Chr.2: 190366886-190366886 on Build GRCh38
rs761618141 Chr.2: 190366881-190366881 on Build GRCh38
rs766586015 Chr.2: 190366855-190366855 on Build GRCh38
rs771459549 Chr.2: 190366841-190366841 on Build GRCh38
rs758183651 Chr.2: 190366854-190366854 on Build GRCh38
rs753860258 Chr.2: 190366901-190366901 on Build GRCh38
rs778168268 Chr.2: 190366832-190366832 on Build GRCh38
rs1028771586 Chr.2: 190367000-190367000 on Build GRCh38
rs780975945 Chr.2: 190366934-190366934 on Build GRCh38
rs775017663 Chr.2: 190366852-190366852 on Build GRCh38
rs767368064 Chr.2: 190366885-190366885 on Build GRCh38
rs192420394 Chr.2: 190366970-190366970 on Build GRCh38
rs976751924 Chr.2: 190366848-190366848 on Build GRCh38
rs765422409 Chr.2: 190366918-190366918 on Build GRCh38
rs200757562 Chr.2: 190366927-190366927 on Build GRCh38
rs918270849 Chr.2: 190366979-190366979 on Build GRCh38
rs537492123 Chr.2: 190366933-190366934 on Build GRCh38
rs754980866 Chr.2: 190366914-190366914 on Build GRCh38
rs547444347 Chr.2: 190367013-190367013 on Build GRCh38
rs751117219 Chr.2: 190366897-190366898 on Build GRCh38
rs111870377 Chr.2: 190366959-190366959 on Build GRCh38
rs543447029 Chr.2: 190366922-190366922 on Build GRCh38
rs766247477 Chr.2: 190366900-190366900 on Build GRCh38

View Full Product Details