Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4783588-4783849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5Z1
Entrez Gene ID
9907
Gene Name
adaptor related protein complex 5 zeta 1 subunit
Gene Aliases
KIAA0415, SPG48, zeta
UniGene
Hs.558440
Gene Chromosome Location
Chr.7: 4775617-4794395 on Build GRCh38
Transcripts
NM_014855.2 XM_017012864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4883122 c.2685G>A p.L895L Chr.7: 4783729-4783729 on Build GRCh38
4502586 c.2749C>T p.R917W Chr.7: 4783793-4783793 on Build GRCh38
1211906 c.2738C>T p.T913M Chr.7: 4783782-4783782 on Build GRCh38
3950565 c.2679C>T p.D893D Chr.7: 4783723-4783723 on Build GRCh38
3950567 c.2754G>C p.Q918H Chr.7: 4783798-4783798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778469154 Chr.7: 4783741-4783741 on Build GRCh38
rs764806337 Chr.7: 4783804-4783804 on Build GRCh38
rs756182519 Chr.7: 4783782-4783782 on Build GRCh38
rs984863672 Chr.7: 4783776-4783776 on Build GRCh38
rs928947081 Chr.7: 4783783-4783783 on Build GRCh38
rs772324497 Chr.7: 4783816-4783816 on Build GRCh38
rs569233326 Chr.7: 4783713-4783713 on Build GRCh38
rs931920386 Chr.7: 4783718-4783718 on Build GRCh38
rs765864595 Chr.7: 4783730-4783730 on Build GRCh38
rs956151272 Chr.7: 4783616-4783616 on Build GRCh38
rs776360426 Chr.7: 4783668-4783669 on Build GRCh38
rs542309905 Chr.7: 4783635-4783635 on Build GRCh38
rs1010349689 Chr.7: 4783756-4783756 on Build GRCh38
rs190350697 Chr.7: 4783789-4783789 on Build GRCh38
rs571423495 Chr.7: 4783661-4783661 on Build GRCh38
rs528229484 Chr.7: 4783650-4783650 on Build GRCh38
rs373829290 Chr.7: 4783727-4783727 on Build GRCh38
rs752495902 Chr.7: 4783752-4783752 on Build GRCh38
rs907834214 Chr.7: 4783681-4783681 on Build GRCh38
rs1044180182 Chr.7: 4783731-4783731 on Build GRCh38
rs759357796 Chr.7: 4783771-4783773 on Build GRCh38
rs546392230 Chr.7: 4783653-4783653 on Build GRCh38
rs930740494 Chr.7: 4783655-4783655 on Build GRCh38
rs769721896 Chr.7: 4783617-4783617 on Build GRCh38
rs750938250 Chr.7: 4783726-4783726 on Build GRCh38
rs1000589136 Chr.7: 4783733-4783733 on Build GRCh38
rs146665638 Chr.7: 4783765-4783765 on Build GRCh38
rs1000029991 Chr.7: 4783736-4783736 on Build GRCh38
rs372609339 Chr.7: 4783668-4783668 on Build GRCh38
rs201030166 Chr.7: 4783643-4783643 on Build GRCh38
rs117659667 Chr.7: 4783761-4783761 on Build GRCh38
rs1040399898 Chr.7: 4783712-4783712 on Build GRCh38
rs920254695 Chr.7: 4783690-4783690 on Build GRCh38
rs1034440470 Chr.7: 4783772-4783772 on Build GRCh38
rs983115035 Chr.7: 4783792-4783792 on Build GRCh38
rs748484751 Chr.7: 4783801-4783801 on Build GRCh38
rs577396839 Chr.7: 4783784-4783784 on Build GRCh38
rs952320860 Chr.7: 4783773-4783773 on Build GRCh38
rs371534878 Chr.7: 4783735-4783735 on Build GRCh38
rs186028694 Chr.7: 4783662-4783662 on Build GRCh38
rs1050248739 Chr.7: 4783720-4783720 on Build GRCh38
rs745887314 Chr.7: 4783656-4783656 on Build GRCh38
rs920304598 Chr.7: 4783779-4783779 on Build GRCh38
rs768017182 Chr.7: 4783717-4783717 on Build GRCh38
rs923307598 Chr.7: 4783827-4783827 on Build GRCh38
rs559118885 Chr.7: 4783771-4783771 on Build GRCh38
rs886062350 Chr.7: 4783762-4783762 on Build GRCh38
rs757322471 Chr.7: 4783769-4783769 on Build GRCh38
rs761451474 Chr.7: 4783793-4783793 on Build GRCh38
rs746939195 Chr.7: 4783766-4783766 on Build GRCh38
rs200456239 Chr.7: 4783767-4783767 on Build GRCh38
rs971643590 Chr.7: 4783628-4783628 on Build GRCh38
rs369252134 Chr.7: 4783807-4783807 on Build GRCh38
rs774461879 Chr.7: 4783794-4783794 on Build GRCh38
rs763255463 Chr.7: 4783665-4783665 on Build GRCh38
rs1029864396 Chr.7: 4783632-4783632 on Build GRCh38
rs556628828 Chr.7: 4783822-4783822 on Build GRCh38
rs754635814 Chr.7: 4783738-4783738 on Build GRCh38
rs759753161 Chr.7: 4783648-4783648 on Build GRCh38
rs762118310 Chr.7: 4783715-4783715 on Build GRCh38
rs73305371 Chr.7: 4783682-4783682 on Build GRCh38

View Full Product Details