Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36871483-36871756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf89
Entrez Gene ID
221477
Gene Name
chromosome 6 open reading frame 89
Gene Aliases
BRAP
UniGene
Hs.433381
Gene Chromosome Location
Chr.6: 36834886-36928964 on Build GRCh38
Transcripts
XM_017010434.1 NM_016059.4

Gene Symbol
PPIL1
Entrez Gene ID
51645
Gene Name
peptidylprolyl isomerase like 1
Gene Aliases
CGI-124, CYPL1, PPIase, hCyPX
UniGene
Hs.27693
Gene Chromosome Location
Chr.6: 36854829-36875024 on Build GRCh38
Transcripts
XM_017010434.1 NM_016059.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3662473 c.211+1G>A p.? Chr.6: 36871717-36871717 on Build GRCh38
7107547 c.175G>C p.D59H Chr.6: 36871754-36871754 on Build GRCh38
5414913 c.211+6G>T p.? Chr.6: 36871712-36871712 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs180912244 Chr.6: 36871617-36871617 on Build GRCh38
rs1017478572 Chr.6: 36871518-36871518 on Build GRCh38
rs35153561 Chr.6: 36871657-36871657 on Build GRCh38
rs920976345 Chr.6: 36871674-36871674 on Build GRCh38
rs371453308 Chr.6: 36871722-36871722 on Build GRCh38
rs374528191 Chr.6: 36871666-36871666 on Build GRCh38
rs748336580 Chr.6: 36871675-36871676 on Build GRCh38
rs752609788 Chr.6: 36871667-36871667 on Build GRCh38
rs76813573 Chr.6: 36871685-36871685 on Build GRCh38
rs749346131 Chr.6: 36871731-36871731 on Build GRCh38
rs906057182 Chr.6: 36871628-36871628 on Build GRCh38
rs976222627 Chr.6: 36871536-36871536 on Build GRCh38
rs566735571 Chr.6: 36871657-36871657 on Build GRCh38
rs909427113 Chr.6: 36871583-36871583 on Build GRCh38
rs779063565 Chr.6: 36871676-36871676 on Build GRCh38
rs768212180 Chr.6: 36871624-36871624 on Build GRCh38
rs575112782 Chr.6: 36871651-36871651 on Build GRCh38
rs529416244 Chr.6: 36871559-36871559 on Build GRCh38
rs772136964 Chr.6: 36871712-36871723 on Build GRCh38
rs780654924 Chr.6: 36871647-36871647 on Build GRCh38
rs372903329 Chr.6: 36871623-36871623 on Build GRCh38
rs543309959 Chr.6: 36871526-36871527 on Build GRCh38
rs185186786 Chr.6: 36871698-36871698 on Build GRCh38
rs551047821 Chr.6: 36871586-36871586 on Build GRCh38
rs929271205 Chr.6: 36871570-36871570 on Build GRCh38
rs143935769 Chr.6: 36871602-36871602 on Build GRCh38
rs756320549 Chr.6: 36871708-36871708 on Build GRCh38
rs982937161 Chr.6: 36871608-36871608 on Build GRCh38
rs779615206 Chr.6: 36871579-36871579 on Build GRCh38
rs114080081 Chr.6: 36871677-36871677 on Build GRCh38
rs9470422 Chr.6: 36871535-36871535 on Build GRCh38
rs777567134 Chr.6: 36871697-36871697 on Build GRCh38

View Full Product Details