Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108552005-108552272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1524
Entrez Gene ID
57650
Gene Name
KIAA1524
Gene Aliases
CIP2A, p90
UniGene
Hs.591308
Gene Chromosome Location
Chr.3: 108545752-108589644 on Build GRCh38
Transcripts
NM_020890.2 XM_006713716.3 XM_006713717.2 XM_011513056.1 XM_011513057.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
580725 c.2538G>C p.L846F Chr.3: 108552243-108552243 on Build GRCh38
71321 c.2518G>A p.E840K Chr.3: 108552263-108552263 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045855050 Chr.3: 108552185-108552185 on Build GRCh38
rs897622697 Chr.3: 108552028-108552028 on Build GRCh38
rs758028835 Chr.3: 108552217-108552217 on Build GRCh38
rs536841356 Chr.3: 108552111-108552111 on Build GRCh38
rs940533409 Chr.3: 108552204-108552204 on Build GRCh38
rs186076795 Chr.3: 108552138-108552138 on Build GRCh38
rs750429462 Chr.3: 108552236-108552236 on Build GRCh38
rs570306943 Chr.3: 108552159-108552159 on Build GRCh38
rs559007011 Chr.3: 108552173-108552173 on Build GRCh38
rs867557238 Chr.3: 108552154-108552154 on Build GRCh38
rs372978208 Chr.3: 108552191-108552191 on Build GRCh38
rs1045009166 Chr.3: 108552065-108552065 on Build GRCh38
rs757058668 Chr.3: 108552225-108552225 on Build GRCh38
rs752375488 Chr.3: 108552201-108552201 on Build GRCh38
rs2305514 Chr.3: 108552165-108552165 on Build GRCh38
rs980467159 Chr.3: 108552108-108552108 on Build GRCh38
rs1036261947 Chr.3: 108552211-108552211 on Build GRCh38
rs763842814 Chr.3: 108552218-108552218 on Build GRCh38
rs977632188 Chr.3: 108552077-108552077 on Build GRCh38
rs369964414 Chr.3: 108552182-108552182 on Build GRCh38
rs3816487 Chr.3: 108552036-108552036 on Build GRCh38
rs572444209 Chr.3: 108552219-108552219 on Build GRCh38
rs375304425 Chr.3: 108552054-108552054 on Build GRCh38
rs1033310809 Chr.3: 108552137-108552137 on Build GRCh38
rs148074605 Chr.3: 108552235-108552235 on Build GRCh38
rs1019489796 Chr.3: 108552034-108552034 on Build GRCh38
rs530571733 Chr.3: 108552087-108552087 on Build GRCh38
rs866227888 Chr.3: 108552055-108552055 on Build GRCh38
rs979622863 Chr.3: 108552240-108552240 on Build GRCh38
rs915921341 Chr.3: 108552181-108552181 on Build GRCh38

View Full Product Details