Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 25526754-25526991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNTD1
Entrez Gene ID
160492
Gene Name
lamin tail domain containing 1
Gene Aliases
IFLTD1, PAS1C1
UniGene
Hs.44647
Gene Chromosome Location
Chr.12: 25476082-25648604 on Build GRCh38
Transcripts
NM_001145727.2 NM_001145728.2 NM_001145729.1 NM_001256266.1 NM_152590.3 XM_011520575.1 XM_011520576.2 XM_011520577.2 XM_011520578.2 XM_011520579.2 XM_011520580.1 XM_011520582.1 XM_011520583.1 XM_011520584.1 XM_011520585.2 XM_011520587.1 XM_011520588.1 XM_017018891.1 XM_017018892.1 XM_017018893.1 XM_017018894.1 XM_017018895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3871315 c.546G>A p.V182V Chr.12: 25526838-25526838 on Build GRCh38
3416731 c.534T>G p.G178G Chr.12: 25526913-25526913 on Build GRCh38
3459519 c.644C>T p.P215L Chr.12: 25526803-25526803 on Build GRCh38
938175 c.532C>T p.L178F Chr.12: 25526852-25526852 on Build GRCh38
3980969 c.508G>C p.E170Q Chr.12: 25526876-25526876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772095431 Chr.12: 25526830-25526830 on Build GRCh38
rs138135357 Chr.12: 25526797-25526797 on Build GRCh38
rs200485290 Chr.12: 25526889-25526889 on Build GRCh38
rs935969750 Chr.12: 25526952-25526952 on Build GRCh38
rs868482390 Chr.12: 25526789-25526789 on Build GRCh38
rs150558545 Chr.12: 25526813-25526813 on Build GRCh38
rs983699601 Chr.12: 25526878-25526878 on Build GRCh38
rs995911346 Chr.12: 25526882-25526882 on Build GRCh38
rs908140513 Chr.12: 25526901-25526901 on Build GRCh38
rs12824916 Chr.12: 25526926-25526926 on Build GRCh38
rs192437605 Chr.12: 25526896-25526896 on Build GRCh38
rs760487363 Chr.12: 25526960-25526960 on Build GRCh38
rs755397657 Chr.12: 25526918-25526918 on Build GRCh38
rs924503756 Chr.12: 25526941-25526941 on Build GRCh38
rs535308374 Chr.12: 25526857-25526857 on Build GRCh38
rs1033721227 Chr.12: 25526900-25526900 on Build GRCh38
rs775434426 Chr.12: 25526866-25526866 on Build GRCh38
rs774211919 Chr.12: 25526898-25526898 on Build GRCh38
rs139058419 Chr.12: 25526914-25526914 on Build GRCh38
rs553205140 Chr.12: 25526812-25526812 on Build GRCh38
rs76122503 Chr.12: 25526810-25526810 on Build GRCh38
rs898773549 Chr.12: 25526863-25526863 on Build GRCh38
rs780421507 Chr.12: 25526799-25526799 on Build GRCh38
rs779372806 Chr.12: 25526919-25526919 on Build GRCh38
rs766664572 Chr.12: 25526913-25526913 on Build GRCh38
rs149733853 Chr.12: 25526795-25526795 on Build GRCh38
rs144446948 Chr.12: 25526910-25526910 on Build GRCh38
rs372692618 Chr.12: 25526796-25526796 on Build GRCh38
rs35628916 Chr.12: 25526907-25526907 on Build GRCh38
rs139538324 Chr.12: 25526906-25526906 on Build GRCh38
rs748249802 Chr.12: 25526821-25526821 on Build GRCh38
rs544334976 Chr.12: 25526858-25526858 on Build GRCh38
rs747224380 Chr.12: 25526847-25526847 on Build GRCh38
rs778028189 Chr.12: 25526835-25526835 on Build GRCh38

View Full Product Details