Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75639494-75639753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMP3
Entrez Gene ID
55272
Gene Name
IMP3, U3 small nucleolar ribonucleoprotein
Gene Aliases
BRMS2, C15orf12, MRPS4
UniGene
Hs.513043
Gene Chromosome Location
Chr.15: 75639085-75640323 on Build GRCh38
Transcripts
NM_018285.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1742118 c.530G>C p.R177P Chr.15: 75639639-75639639 on Build GRCh38
1290433 c.433G>C p.D145H Chr.15: 75639736-75639736 on Build GRCh38
4692428 c.517T>C p.Y173H Chr.15: 75639652-75639652 on Build GRCh38
1749319 c.484G>A p.D162N Chr.15: 75639685-75639685 on Build GRCh38
6685595 c.522T>C p.N174N Chr.15: 75639647-75639647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553887402 Chr.15: 75639533-75639533 on Build GRCh38
rs375180548 Chr.15: 75639659-75639659 on Build GRCh38
rs750844720 Chr.15: 75639686-75639687 on Build GRCh38
rs981260429 Chr.15: 75639545-75639545 on Build GRCh38
rs747709800 Chr.15: 75639585-75639585 on Build GRCh38
rs750879920 Chr.15: 75639671-75639671 on Build GRCh38
rs145101512 Chr.15: 75639615-75639615 on Build GRCh38
rs769839942 Chr.15: 75639726-75639726 on Build GRCh38
rs532458855 Chr.15: 75639729-75639729 on Build GRCh38
rs746754233 Chr.15: 75639618-75639618 on Build GRCh38
rs768043485 Chr.15: 75639566-75639566 on Build GRCh38
rs777193241 Chr.15: 75639708-75639708 on Build GRCh38
rs753510450 Chr.15: 75639568-75639568 on Build GRCh38
rs866193683 Chr.15: 75639709-75639709 on Build GRCh38
rs1006119357 Chr.15: 75639531-75639531 on Build GRCh38
rs1041614779 Chr.15: 75639642-75639642 on Build GRCh38
rs757945550 Chr.15: 75639693-75639693 on Build GRCh38
rs947356387 Chr.15: 75639622-75639622 on Build GRCh38
rs754532383 Chr.15: 75639673-75639673 on Build GRCh38
rs752105243 Chr.15: 75639685-75639685 on Build GRCh38
rs370084628 Chr.15: 75639575-75639575 on Build GRCh38
rs746913670 Chr.15: 75639711-75639711 on Build GRCh38
rs757251077 Chr.15: 75639715-75639715 on Build GRCh38
rs904889187 Chr.15: 75639588-75639588 on Build GRCh38
rs374444374 Chr.15: 75639580-75639580 on Build GRCh38
rs770111406 Chr.15: 75639647-75639647 on Build GRCh38
rs758206268 Chr.15: 75639603-75639603 on Build GRCh38
rs762451717 Chr.15: 75639661-75639661 on Build GRCh38
rs370385015 Chr.15: 75639638-75639638 on Build GRCh38
rs141574567 Chr.15: 75639628-75639628 on Build GRCh38
rs763126799 Chr.15: 75639649-75639649 on Build GRCh38
rs780951310 Chr.15: 75639717-75639717 on Build GRCh38
rs745675077 Chr.15: 75639724-75639724 on Build GRCh38
rs956352865 Chr.15: 75639561-75639561 on Build GRCh38
rs767994018 Chr.15: 75639668-75639668 on Build GRCh38
rs148358312 Chr.15: 75639625-75639625 on Build GRCh38
rs200453864 Chr.15: 75639648-75639648 on Build GRCh38
rs765821833 Chr.15: 75639683-75639684 on Build GRCh38
rs762052741 Chr.15: 75639587-75639590 on Build GRCh38
rs754749378 Chr.15: 75639572-75639572 on Build GRCh38
rs762554499 Chr.15: 75639730-75639730 on Build GRCh38
rs373700434 Chr.15: 75639542-75639542 on Build GRCh38
rs145702963 Chr.15: 75639560-75639560 on Build GRCh38
rs764950244 Chr.15: 75639677-75639677 on Build GRCh38
rs759137053 Chr.15: 75639635-75639635 on Build GRCh38
rs764461683 Chr.15: 75639653-75639653 on Build GRCh38
rs777043428 Chr.15: 75639568-75639569 on Build GRCh38

View Full Product Details