Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 131277078-131277303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF1
Entrez Gene ID
3547
Gene Name
immunoglobulin superfamily member 1
Gene Aliases
CHTE, IGCD1, IGDC1, INHBP, PGSF2, p120
UniGene
Hs.22111
Gene Chromosome Location
Chr.X: 131273506-131289457 on Build GRCh38
Transcripts
NM_001170961.1 NM_001170962.1 NM_001555.4 XM_011531330.1 XM_011531333.1 XM_011531334.1

SNP Detail

SNP ID Chromosome Location
rs774344485 Chr.X: 131277236-131277237 on Build GRCh38
rs747955793 Chr.X: 131277266-131277266 on Build GRCh38
rs369042210 Chr.X: 131277133-131277133 on Build GRCh38
rs772067953 Chr.X: 131277254-131277255 on Build GRCh38
rs751748646 Chr.X: 131277184-131277184 on Build GRCh38
rs887942207 Chr.X: 131277198-131277198 on Build GRCh38
rs781166601 Chr.X: 131277129-131277129 on Build GRCh38
rs754947994 Chr.X: 131277254-131277254 on Build GRCh38
rs745912627 Chr.X: 131277253-131277253 on Build GRCh38
rs12393480 Chr.X: 131277190-131277190 on Build GRCh38
rs189592440 Chr.X: 131277248-131277248 on Build GRCh38
rs753155514 Chr.X: 131277134-131277134 on Build GRCh38
rs898047422 Chr.X: 131277261-131277261 on Build GRCh38
rs756134105 Chr.X: 131277256-131277256 on Build GRCh38
rs776004054 Chr.X: 131277108-131277108 on Build GRCh38
rs773228853 Chr.X: 131277257-131277261 on Build GRCh38
rs192379880 Chr.X: 131277268-131277268 on Build GRCh38
rs768805436 Chr.X: 131277237-131277242 on Build GRCh38
rs757289487 Chr.X: 131277230-131277230 on Build GRCh38
rs758796307 Chr.X: 131277138-131277138 on Build GRCh38
rs764245106 Chr.X: 131277164-131277164 on Build GRCh38
rs747969293 Chr.X: 131277248-131277249 on Build GRCh38

View Full Product Details