Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139371283-139371548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf55-LUC7L2
Entrez Gene ID
100996928
Gene Name
C7orf55-LUC7L2 readthrough
Gene Aliases
-
UniGene
Hs.731488
Gene Chromosome Location
Chr.7: 139341132-139423457 on Build GRCh38
Transcripts
NM_001244584.2 NM_001244585.1 NM_001270643.1 NM_016019.4

Gene Symbol
LUC7L2
Entrez Gene ID
51631
Gene Name
LUC7 like 2, pre-mRNA splicing factor
Gene Aliases
CGI-59, CGI-74, LUC7B2
UniGene
Hs.731488
Gene Chromosome Location
Chr.7: 139340359-139423457 on Build GRCh38
Transcripts
NM_001244584.2 NM_001244585.1 NM_001270643.1 NM_016019.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5500440 c.52+10A>C p.? Chr.7: 139371473-139371473 on Build GRCh38
7326117 c.12C>T p.H4H Chr.7: 139371423-139371423 on Build GRCh38
3686356 c.22delA p.I10fs*48 Chr.7: 139371433-139371433 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs940523366 Chr.7: 139371369-139371369 on Build GRCh38
rs530345609 Chr.7: 139371343-139371343 on Build GRCh38
rs538384467 Chr.7: 139371342-139371342 on Build GRCh38
rs923400534 Chr.7: 139371424-139371424 on Build GRCh38
rs772488102 Chr.7: 139371432-139371433 on Build GRCh38
rs992174544 Chr.7: 139371515-139371515 on Build GRCh38
rs972281339 Chr.7: 139371370-139371370 on Build GRCh38
rs553855864 Chr.7: 139371506-139371506 on Build GRCh38
rs542179637 Chr.7: 139371423-139371423 on Build GRCh38
rs758644557 Chr.7: 139371518-139371518 on Build GRCh38
rs951443644 Chr.7: 139371402-139371402 on Build GRCh38
rs544411117 Chr.7: 139371318-139371318 on Build GRCh38
rs933364033 Chr.7: 139371426-139371426 on Build GRCh38
rs1050427467 Chr.7: 139371477-139371477 on Build GRCh38
rs140959718 Chr.7: 139371338-139371338 on Build GRCh38
rs889115124 Chr.7: 139371497-139371497 on Build GRCh38

View Full Product Details