Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32908319-32908576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB1
Entrez Gene ID
3688
Gene Name
integrin subunit beta 1
Gene Aliases
CD29, FNRB, GPIIA, MDF2, MSK12, VLA-BETA, VLAB
UniGene
Hs.643813
Gene Chromosome Location
Chr.10: 32900318-32958365 on Build GRCh38
Transcripts
NM_002211.3 NM_033668.2 NM_133376.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5104559 c.2228T>C p.I743T Chr.10: 32908471-32908471 on Build GRCh38
5359878 c.2331+3A>T p.? Chr.10: 32908365-32908365 on Build GRCh38
6367894 c.2234T>A p.L745H Chr.10: 32908465-32908465 on Build GRCh38
4974828 c.2276A>T p.D759V Chr.10: 32908423-32908423 on Build GRCh38
4957330 c.2273A>G p.H758R Chr.10: 32908426-32908426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201244255 Chr.10: 32908548-32908548 on Build GRCh38
rs749133019 Chr.10: 32908512-32908512 on Build GRCh38
rs779312239 Chr.10: 32908522-32908522 on Build GRCh38
rs775491212 Chr.10: 32908547-32908547 on Build GRCh38
rs764506216 Chr.10: 32908363-32908363 on Build GRCh38
rs754514061 Chr.10: 32908467-32908467 on Build GRCh38
rs760891308 Chr.10: 32908359-32908359 on Build GRCh38
rs201738777 Chr.10: 32908505-32908505 on Build GRCh38
rs879013371 Chr.10: 32908491-32908491 on Build GRCh38
rs1013483956 Chr.10: 32908443-32908443 on Build GRCh38
rs754225572 Chr.10: 32908368-32908368 on Build GRCh38
rs200765922 Chr.10: 32908373-32908373 on Build GRCh38
rs771232092 Chr.10: 32908514-32908514 on Build GRCh38
rs780923698 Chr.10: 32908488-32908488 on Build GRCh38
rs752690214 Chr.10: 32908497-32908497 on Build GRCh38
rs780953431 Chr.10: 32908351-32908352 on Build GRCh38
rs745881141 Chr.10: 32908526-32908526 on Build GRCh38
rs766068569 Chr.10: 32908433-32908433 on Build GRCh38
rs942464178 Chr.10: 32908542-32908542 on Build GRCh38
rs762563378 Chr.10: 32908425-32908425 on Build GRCh38
rs756060620 Chr.10: 32908501-32908501 on Build GRCh38
rs751018992 Chr.10: 32908458-32908458 on Build GRCh38
rs896505656 Chr.10: 32908434-32908434 on Build GRCh38
rs200099735 Chr.10: 32908529-32908529 on Build GRCh38

View Full Product Details