Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121415948-121416221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2B
Entrez Gene ID
84678
Gene Name
lysine demethylase 2B
Gene Aliases
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2
UniGene
Hs.524800
Gene Chromosome Location
Chr.12: 121402289-121582268 on Build GRCh38
Transcripts
XM_011538867.2 NM_001256858.1 NM_025126.3 NM_194271.2

Gene Symbol
RNF34
Entrez Gene ID
80196
Gene Name
ring finger protein 34
Gene Aliases
CARP-1, CARP1, RFI, RIF, RIFF, hRFI
UniGene
Hs.292804
Gene Chromosome Location
Chr.12: 121400083-121424353 on Build GRCh38
Transcripts
XM_011538867.2 NM_001256858.1 NM_025126.3 NM_194271.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1476250 c.18G>A p.T6T Chr.12: 121416170-121416170 on Build GRCh38
6520757 c.51G>A p.L17L Chr.12: 121416200-121416200 on Build GRCh38
1605666 c.45G>A p.L15L Chr.12: 121416197-121416197 on Build GRCh38
5550243 c.17C>T p.A6V Chr.12: 121416166-121416166 on Build GRCh38
1605665 c.48G>A p.L16L Chr.12: 121416197-121416197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539486553 Chr.12: 121416053-121416053 on Build GRCh38
rs782326655 Chr.12: 121416147-121416147 on Build GRCh38
rs1032446702 Chr.12: 121416071-121416071 on Build GRCh38
rs782816324 Chr.12: 121416195-121416195 on Build GRCh38
rs557821046 Chr.12: 121416085-121416085 on Build GRCh38
rs753331968 Chr.12: 121416079-121416079 on Build GRCh38
rs782178280 Chr.12: 121416139-121416139 on Build GRCh38
rs537636270 Chr.12: 121416170-121416170 on Build GRCh38
rs568859609 Chr.12: 121416089-121416089 on Build GRCh38
rs782099773 Chr.12: 121416161-121416161 on Build GRCh38
rs781926683 Chr.12: 121416190-121416190 on Build GRCh38
rs782306563 Chr.12: 121416189-121416189 on Build GRCh38
rs966913315 Chr.12: 121416028-121416028 on Build GRCh38
rs782662202 Chr.12: 121416159-121416159 on Build GRCh38
rs781929620 Chr.12: 121416077-121416077 on Build GRCh38
rs566045979 Chr.12: 121415976-121415976 on Build GRCh38
rs201536085 Chr.12: 121416124-121416124 on Build GRCh38
rs781945749 Chr.12: 121416175-121416175 on Build GRCh38
rs782631536 Chr.12: 121416130-121416130 on Build GRCh38
rs1021618367 Chr.12: 121416008-121416008 on Build GRCh38
rs781851364 Chr.12: 121416140-121416140 on Build GRCh38
rs201929209 Chr.12: 121416185-121416185 on Build GRCh38
rs782139970 Chr.12: 121416194-121416194 on Build GRCh38

View Full Product Details