Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 37129970-37130235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD30A
Entrez Gene ID
91074
Gene Name
ankyrin repeat domain 30A
Gene Aliases
NY-BR-1
UniGene
Hs.373787
Gene Chromosome Location
Chr.10: 37125754-37235831 on Build GRCh38
Transcripts
NM_052997.2 XM_011519757.2 XM_011519758.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6331121 c.148C>A p.H50N Chr.10: 37129987-37129987 on Build GRCh38
5613208 c.158C>A p.P53H Chr.10: 37129997-37129997 on Build GRCh38
159078 c.165G>T p.M55I Chr.10: 37130004-37130004 on Build GRCh38
5800823 c.144C>G p.G48G Chr.10: 37129983-37129983 on Build GRCh38
5800824 c.144C>G p.G48G Chr.10: 37129983-37129983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771592739 Chr.10: 37130030-37130030 on Build GRCh38
rs772166309 Chr.10: 37130059-37130059 on Build GRCh38
rs779748317 Chr.10: 37130198-37130198 on Build GRCh38
rs576006073 Chr.10: 37130084-37130084 on Build GRCh38
rs572452768 Chr.10: 37130106-37130106 on Build GRCh38
rs186223474 Chr.10: 37130161-37130161 on Build GRCh38
rs201813483 Chr.10: 37130200-37130200 on Build GRCh38
rs376005400 Chr.10: 37130001-37130001 on Build GRCh38
rs556109708 Chr.10: 37130081-37130081 on Build GRCh38
rs559007406 Chr.10: 37130092-37130092 on Build GRCh38
rs541574445 Chr.10: 37130113-37130113 on Build GRCh38
rs899935311 Chr.10: 37129998-37129998 on Build GRCh38
rs775167284 Chr.10: 37130155-37130155 on Build GRCh38
rs779644783 Chr.10: 37130180-37130180 on Build GRCh38
rs561339551 Chr.10: 37130124-37130124 on Build GRCh38
rs765213450 Chr.10: 37130185-37130185 on Build GRCh38
rs1004127100 Chr.10: 37130109-37130109 on Build GRCh38
rs11011046 Chr.10: 37130162-37130162 on Build GRCh38
rs781356359 Chr.10: 37130216-37130216 on Build GRCh38
rs568301150 Chr.10: 37129989-37129989 on Build GRCh38
rs751652945 Chr.10: 37130189-37130189 on Build GRCh38
rs755171334 Chr.10: 37130206-37130206 on Build GRCh38
rs1015455653 Chr.10: 37130134-37130134 on Build GRCh38
rs555401398 Chr.10: 37130090-37130090 on Build GRCh38
rs763534436 Chr.10: 37130173-37130173 on Build GRCh38
rs763433088 Chr.10: 37130009-37130009 on Build GRCh38
rs773332296 Chr.10: 37130172-37130183 on Build GRCh38
rs748155942 Chr.10: 37130013-37130015 on Build GRCh38

View Full Product Details