Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4968795-4969062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMTA2
Entrez Gene ID
23125
Gene Name
calmodulin binding transcription activator 2
Gene Aliases
-
UniGene
Hs.632242
Gene Chromosome Location
Chr.17: 4967992-4988297 on Build GRCh38
Transcripts
NM_001171166.1 NM_001171167.1 NM_001171168.1 NM_015099.3 XM_006721478.3 XM_006721481.3 XM_006721482.3 XM_011523746.2 XM_011523747.2 XM_011523748.2 XM_011523749.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4657370 c.3564G>C p.Q1188H Chr.17: 4968801-4968801 on Build GRCh38
3192753 c.3533G>A p.R1178H Chr.17: 4968919-4968919 on Build GRCh38
473027 c.3471-1G>A p.? Chr.17: 4968982-4968982 on Build GRCh38
4268739 c.3600G>A p.A1200A Chr.17: 4968919-4968919 on Build GRCh38
6619628 c.3626A>G p.E1209G Chr.17: 4968806-4968806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750026430 Chr.17: 4968830-4968830 on Build GRCh38
rs747840253 Chr.17: 4968914-4968914 on Build GRCh38
rs756669268 Chr.17: 4969018-4969018 on Build GRCh38
rs563053214 Chr.17: 4968944-4968944 on Build GRCh38
rs886944532 Chr.17: 4968869-4968869 on Build GRCh38
rs1001058184 Chr.17: 4968892-4968892 on Build GRCh38
rs374538074 Chr.17: 4969022-4969022 on Build GRCh38
rs992401556 Chr.17: 4968821-4968821 on Build GRCh38
rs750151051 Chr.17: 4968956-4968956 on Build GRCh38
rs762869651 Chr.17: 4968961-4968961 on Build GRCh38
rs753208472 Chr.17: 4969013-4969013 on Build GRCh38
rs751024487 Chr.17: 4968860-4968860 on Build GRCh38
rs777126758 Chr.17: 4969014-4969014 on Build GRCh38
rs759524981 Chr.17: 4968913-4968913 on Build GRCh38
rs753835192 Chr.17: 4968818-4968818 on Build GRCh38
rs750580371 Chr.17: 4968827-4968828 on Build GRCh38
rs751237076 Chr.17: 4969004-4969004 on Build GRCh38
rs200440540 Chr.17: 4968835-4968835 on Build GRCh38
rs751364062 Chr.17: 4968856-4968856 on Build GRCh38
rs754377318 Chr.17: 4969007-4969007 on Build GRCh38
rs779792729 Chr.17: 4968856-4968856 on Build GRCh38
rs758689311 Chr.17: 4968836-4968838 on Build GRCh38
rs765934356 Chr.17: 4968973-4968973 on Build GRCh38
rs780947854 Chr.17: 4969033-4969034 on Build GRCh38
rs1042899112 Chr.17: 4969038-4969038 on Build GRCh38
rs1044847064 Chr.17: 4968817-4968817 on Build GRCh38
rs748116223 Chr.17: 4969034-4969034 on Build GRCh38
rs748859332 Chr.17: 4969040-4969040 on Build GRCh38
rs1053084715 Chr.17: 4968995-4968995 on Build GRCh38
rs370852194 Chr.17: 4968870-4968870 on Build GRCh38
rs770242409 Chr.17: 4968896-4968896 on Build GRCh38
rs765205165 Chr.17: 4968955-4968955 on Build GRCh38
rs371011585 Chr.17: 4969011-4969011 on Build GRCh38
rs1009808855 Chr.17: 4968906-4968906 on Build GRCh38
rs754574318 Chr.17: 4968866-4968866 on Build GRCh38
rs192180588 Chr.17: 4968922-4968922 on Build GRCh38
rs749711823 Chr.17: 4968900-4968900 on Build GRCh38
rs766187363 Chr.17: 4968837-4968838 on Build GRCh38
rs765023306 Chr.17: 4968828-4968828 on Build GRCh38
rs373647680 Chr.17: 4968894-4968894 on Build GRCh38
rs774701224 Chr.17: 4968907-4968907 on Build GRCh38
rs763937985 Chr.17: 4968816-4968816 on Build GRCh38
rs145165015 Chr.17: 4968897-4968897 on Build GRCh38
rs138928424 Chr.17: 4969027-4969027 on Build GRCh38
rs377542650 Chr.17: 4968822-4968822 on Build GRCh38
rs757961464 Chr.17: 4968840-4968840 on Build GRCh38
rs761771999 Chr.17: 4968952-4968952 on Build GRCh38
rs763552126 Chr.17: 4968887-4968887 on Build GRCh38

View Full Product Details