Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28830965-28831196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN2L
Entrez Gene ID
11273
Gene Name
ataxin 2 like
Gene Aliases
A2D, A2LG, A2LP, A2RP
UniGene
Hs.460499
Gene Chromosome Location
Chr.16: 28823048-28837237 on Build GRCh38
Transcripts
NM_001308230.1 NM_007245.3 NM_017492.3 NM_145714.2 NM_148414.2 NM_148415.2 NM_148416.2 XM_005255061.1 XM_005255062.1 XM_005255063.3 XM_005255064.1 XM_005255065.1 XM_005255066.1 XM_005255067.1 XM_005255068.1 XM_005255069.1 XM_005255070.1 XM_005255071.1 XM_005255074.1 XM_005255075.1 XM_005255076.1 XM_005255077.1 XM_006721007.1 XM_006721008.1 XM_006721009.1 XM_006721010.2 XM_006721011.1 XM_006721012.3 XM_011545719.2 XM_011545721.2 XM_011545722.1 XM_017022891.1 XM_017022892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6608704 c.1271C>T p.S424L Chr.16: 28831022-28831022 on Build GRCh38
6282024 c.1217C>A p.S406Y Chr.16: 28830968-28830968 on Build GRCh38
4664678 c.1315delC p.P439fs*42 Chr.16: 28831066-28831066 on Build GRCh38
4664673 c.1272G>A p.S424S Chr.16: 28831023-28831023 on Build GRCh38
4804371 c.1259A>T p.K420M Chr.16: 28831010-28831010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567644246 Chr.16: 28831136-28831136 on Build GRCh38
rs755296971 Chr.16: 28831063-28831063 on Build GRCh38
rs766756824 Chr.16: 28831023-28831023 on Build GRCh38
rs552074799 Chr.16: 28831017-28831017 on Build GRCh38
rs763379472 Chr.16: 28831022-28831022 on Build GRCh38
rs749814513 Chr.16: 28831096-28831096 on Build GRCh38
rs371862735 Chr.16: 28831052-28831052 on Build GRCh38
rs778821255 Chr.16: 28831057-28831057 on Build GRCh38
rs750652233 Chr.16: 28831096-28831100 on Build GRCh38
rs759814662 Chr.16: 28831103-28831103 on Build GRCh38
rs12597149 Chr.16: 28831112-28831112 on Build GRCh38
rs748339852 Chr.16: 28831069-28831069 on Build GRCh38
rs781516862 Chr.16: 28831053-28831053 on Build GRCh38
rs776813801 Chr.16: 28831008-28831008 on Build GRCh38
rs910386353 Chr.16: 28831128-28831128 on Build GRCh38
rs755700917 Chr.16: 28831049-28831049 on Build GRCh38
rs979640287 Chr.16: 28831122-28831122 on Build GRCh38
rs745622824 Chr.16: 28831062-28831062 on Build GRCh38
rs748175093 Chr.16: 28831010-28831010 on Build GRCh38
rs199863879 Chr.16: 28831041-28831041 on Build GRCh38
rs908531780 Chr.16: 28831132-28831132 on Build GRCh38
rs767256529 Chr.16: 28831104-28831104 on Build GRCh38
rs975795002 Chr.16: 28831158-28831158 on Build GRCh38
rs774529844 Chr.16: 28831028-28831028 on Build GRCh38
rs767044281 Chr.16: 28831033-28831033 on Build GRCh38
rs146367102 Chr.16: 28831066-28831066 on Build GRCh38
rs369871413 Chr.16: 28831093-28831093 on Build GRCh38
rs368508890 Chr.16: 28831046-28831046 on Build GRCh38
rs373923639 Chr.16: 28831034-28831034 on Build GRCh38
rs4344749 Chr.16: 28830990-28830990 on Build GRCh38
rs371700809 Chr.16: 28831013-28831013 on Build GRCh38
rs778811567 Chr.16: 28830992-28830992 on Build GRCh38
rs774817399 Chr.16: 28831102-28831102 on Build GRCh38
rs759281381 Chr.16: 28831031-28831031 on Build GRCh38
rs769864423 Chr.16: 28831084-28831084 on Build GRCh38
rs775010366 Chr.16: 28831110-28831110 on Build GRCh38
rs1047089796 Chr.16: 28831123-28831123 on Build GRCh38
rs923051216 Chr.16: 28831163-28831163 on Build GRCh38
rs768743112 Chr.16: 28831006-28831006 on Build GRCh38
rs150743711 Chr.16: 28831064-28831064 on Build GRCh38
rs943243336 Chr.16: 28831131-28831131 on Build GRCh38
rs1009258367 Chr.16: 28831039-28831039 on Build GRCh38
rs371752882 Chr.16: 28831099-28831099 on Build GRCh38
rs1041072398 Chr.16: 28831145-28831145 on Build GRCh38
rs532048463 Chr.16: 28830993-28830993 on Build GRCh38

View Full Product Details