Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 82354306-82354545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC59
Entrez Gene ID
29080
Gene Name
coiled-coil domain containing 59
Gene Aliases
BR22, HSPC128, TAP26
UniGene
Hs.582627
Gene Chromosome Location
Chr.12: 82352304-82358805 on Build GRCh38
Transcripts
NM_014167.4 NR_033192.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5000371 c.561_564+4delACAAGTAA p.? Chr.12: 82354491-82354498 on Build GRCh38
4957312 c.560A>C p.K187T Chr.12: 82354499-82354499 on Build GRCh38
1993764 c.548G>A p.R183H Chr.12: 82354511-82354511 on Build GRCh38
145201 c.538C>A p.Q180K Chr.12: 82354521-82354521 on Build GRCh38
943952 c.547C>T p.R183C Chr.12: 82354512-82354512 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763696039 Chr.12: 82354481-82354481 on Build GRCh38
rs778712677 Chr.12: 82354410-82354410 on Build GRCh38
rs867173016 Chr.12: 82354388-82354390 on Build GRCh38
rs758917272 Chr.12: 82354511-82354511 on Build GRCh38
rs1056289303 Chr.12: 82354339-82354339 on Build GRCh38
rs750969222 Chr.12: 82354508-82354508 on Build GRCh38
rs186194036 Chr.12: 82354435-82354435 on Build GRCh38
rs756144512 Chr.12: 82354489-82354492 on Build GRCh38
rs1027968358 Chr.12: 82354400-82354400 on Build GRCh38
rs192072744 Chr.12: 82354514-82354514 on Build GRCh38
rs527805247 Chr.12: 82354474-82354474 on Build GRCh38
rs777170961 Chr.12: 82354345-82354345 on Build GRCh38
rs1043462363 Chr.12: 82354367-82354367 on Build GRCh38
rs551829030 Chr.12: 82354365-82354365 on Build GRCh38
rs749125579 Chr.12: 82354495-82354498 on Build GRCh38
rs753508111 Chr.12: 82354497-82354497 on Build GRCh38
rs754862285 Chr.12: 82354356-82354356 on Build GRCh38
rs150615186 Chr.12: 82354512-82354512 on Build GRCh38
rs368708325 Chr.12: 82354389-82354390 on Build GRCh38
rs759404900 Chr.12: 82354447-82354447 on Build GRCh38
rs969884826 Chr.12: 82354400-82354401 on Build GRCh38
rs537485997 Chr.12: 82354491-82354498 on Build GRCh38
rs374928926 Chr.12: 82354516-82354516 on Build GRCh38
rs566858842 Chr.12: 82354397-82354397 on Build GRCh38
rs1008908667 Chr.12: 82354427-82354427 on Build GRCh38
rs533676618 Chr.12: 82354336-82354336 on Build GRCh38
rs375035875 Chr.12: 82354467-82354467 on Build GRCh38
rs996464430 Chr.12: 82354392-82354392 on Build GRCh38
rs371315969 Chr.12: 82354448-82354448 on Build GRCh38
rs372398424 Chr.12: 82354478-82354478 on Build GRCh38
rs903645406 Chr.12: 82354389-82354389 on Build GRCh38
rs148916685 Chr.12: 82354388-82354389 on Build GRCh38
rs762140623 Chr.12: 82354503-82354503 on Build GRCh38
rs765690739 Chr.12: 82354507-82354507 on Build GRCh38
rs892310348 Chr.12: 82354341-82354341 on Build GRCh38

View Full Product Details