Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3917641-3917894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATCAY
Entrez Gene ID
85300
Gene Name
ATCAY, caytaxin
Gene Aliases
BNIP-H, CLAC
UniGene
Hs.418055
Gene Chromosome Location
Chr.19: 3880620-3928082 on Build GRCh38
Transcripts
NM_033064.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5623858 c.980G>C p.R327T Chr.19: 3917756-3917756 on Build GRCh38
3422779 c.969C>T p.Y323Y Chr.19: 3917745-3917745 on Build GRCh38
5623857 c.980G>C p.R327T Chr.19: 3917756-3917756 on Build GRCh38
6151193 c.997G>C p.E333Q Chr.19: 3917773-3917773 on Build GRCh38
3422778 c.969C>T p.Y323Y Chr.19: 3917745-3917745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989818780 Chr.19: 3917828-3917828 on Build GRCh38
rs773654978 Chr.19: 3917825-3917825 on Build GRCh38
rs761094085 Chr.19: 3917819-3917820 on Build GRCh38
rs750440597 Chr.19: 3917800-3917800 on Build GRCh38
rs914555853 Chr.19: 3917829-3917829 on Build GRCh38
rs947725462 Chr.19: 3917768-3917768 on Build GRCh38
rs927883648 Chr.19: 3917807-3917807 on Build GRCh38
rs371775210 Chr.19: 3917816-3917816 on Build GRCh38
rs374690024 Chr.19: 3917690-3917690 on Build GRCh38
rs770255637 Chr.19: 3917701-3917701 on Build GRCh38
rs897223626 Chr.19: 3917838-3917838 on Build GRCh38
rs368207414 Chr.19: 3917809-3917809 on Build GRCh38
rs989099680 Chr.19: 3917743-3917743 on Build GRCh38
rs561750458 Chr.19: 3917789-3917789 on Build GRCh38
rs372049992 Chr.19: 3917762-3917762 on Build GRCh38
rs923696563 Chr.19: 3917673-3917673 on Build GRCh38
rs978685314 Chr.19: 3917668-3917668 on Build GRCh38
rs757155455 Chr.19: 3917765-3917765 on Build GRCh38
rs546231423 Chr.19: 3917841-3917844 on Build GRCh38
rs773741130 Chr.19: 3917711-3917711 on Build GRCh38
rs368626484 Chr.19: 3917751-3917751 on Build GRCh38
rs763043470 Chr.19: 3917717-3917717 on Build GRCh38
rs776967741 Chr.19: 3917692-3917692 on Build GRCh38
rs940510067 Chr.19: 3917811-3917811 on Build GRCh38
rs780865061 Chr.19: 3917820-3917820 on Build GRCh38
rs759668217 Chr.19: 3917746-3917746 on Build GRCh38
rs766386513 Chr.19: 3917722-3917722 on Build GRCh38
rs770169410 Chr.19: 3917823-3917823 on Build GRCh38
rs971763258 Chr.19: 3917724-3917724 on Build GRCh38
rs376021315 Chr.19: 3917819-3917819 on Build GRCh38
rs751742218 Chr.19: 3917745-3917745 on Build GRCh38
rs779434825 Chr.19: 3917812-3917812 on Build GRCh38
rs374312343 Chr.19: 3917803-3917803 on Build GRCh38
rs762215733 Chr.19: 3917697-3917697 on Build GRCh38

View Full Product Details