Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89279302-89279551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
UniGene
Hs.335003
Gene Chromosome Location
Chr.16: 89267619-89490561 on Build GRCh38
Transcripts
NM_001256182.1 NM_001256183.1 NM_013275.5 XM_006721181.1 XM_006721184.2 XM_011523051.2 XM_011523053.2 XM_011523054.2 XM_011523055.2 XM_011523056.2 XM_011523057.2 XM_017023182.1 XM_017023183.1 XM_017023184.1 XM_017023185.1 XM_017023186.1 XM_017023187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5427810 c.7022C>T p.A2341V Chr.16: 89279520-89279520 on Build GRCh38
3932466 c.7106C>T p.A2369V Chr.16: 89279436-89279436 on Build GRCh38
4129516 c.7128C>G p.D2376E Chr.16: 89279414-89279414 on Build GRCh38
4617711 c.7190A>T p.Q2397L Chr.16: 89279352-89279352 on Build GRCh38
292711 c.7131C>T p.D2377D Chr.16: 89279411-89279411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759740258 Chr.16: 89279344-89279344 on Build GRCh38
rs773056661 Chr.16: 89279330-89279330 on Build GRCh38
rs763041579 Chr.16: 89279332-89279332 on Build GRCh38
rs761715813 Chr.16: 89279321-89279321 on Build GRCh38
rs779428707 Chr.16: 89279381-89279381 on Build GRCh38
rs774236725 Chr.16: 89279507-89279507 on Build GRCh38
rs548304688 Chr.16: 89279360-89279360 on Build GRCh38
rs1006297836 Chr.16: 89279525-89279525 on Build GRCh38
rs1036666745 Chr.16: 89279410-89279410 on Build GRCh38
rs1024160802 Chr.16: 89279496-89279496 on Build GRCh38
rs533765590 Chr.16: 89279510-89279510 on Build GRCh38
rs4031012 Chr.16: 89279520-89279520 on Build GRCh38
rs4785560 Chr.16: 89279414-89279414 on Build GRCh38
rs927582667 Chr.16: 89279375-89279375 on Build GRCh38
rs863225296 Chr.16: 89279362-89279362 on Build GRCh38
rs749176560 Chr.16: 89279481-89279481 on Build GRCh38
rs753291494 Chr.16: 89279407-89279409 on Build GRCh38
rs1018954790 Chr.16: 89279439-89279439 on Build GRCh38
rs747911603 Chr.16: 89279431-89279431 on Build GRCh38
rs769501453 Chr.16: 89279455-89279455 on Build GRCh38
rs770957459 Chr.16: 89279501-89279501 on Build GRCh38
rs535866230 Chr.16: 89279358-89279358 on Build GRCh38
rs756497363 Chr.16: 89279352-89279352 on Build GRCh38
rs889244275 Chr.16: 89279426-89279426 on Build GRCh38
rs886041822 Chr.16: 89279458-89279459 on Build GRCh38
rs550312255 Chr.16: 89279336-89279336 on Build GRCh38
rs991655950 Chr.16: 89279508-89279508 on Build GRCh38
rs756783131 Chr.16: 89279414-89279415 on Build GRCh38
rs994703276 Chr.16: 89279413-89279413 on Build GRCh38
rs999130123 Chr.16: 89279452-89279452 on Build GRCh38
rs947802091 Chr.16: 89279340-89279340 on Build GRCh38
rs986291618 Chr.16: 89279401-89279401 on Build GRCh38
rs867650813 Chr.16: 89279484-89279484 on Build GRCh38
rs767682539 Chr.16: 89279528-89279528 on Build GRCh38
rs773110185 Chr.16: 89279471-89279471 on Build GRCh38
rs767739621 Chr.16: 89279351-89279351 on Build GRCh38
rs899013747 Chr.16: 89279411-89279411 on Build GRCh38
rs377374681 Chr.16: 89279356-89279356 on Build GRCh38
rs886041942 Chr.16: 89279480-89279480 on Build GRCh38
rs904015120 Chr.16: 89279437-89279437 on Build GRCh38
rs957598792 Chr.16: 89279482-89279482 on Build GRCh38
rs746579362 Chr.16: 89279409-89279409 on Build GRCh38
rs374714506 Chr.16: 89279335-89279335 on Build GRCh38
rs940623226 Chr.16: 89279406-89279406 on Build GRCh38
rs969115786 Chr.16: 89279515-89279515 on Build GRCh38
rs1013550925 Chr.16: 89279487-89279487 on Build GRCh38
rs780996696 Chr.16: 89279427-89279427 on Build GRCh38
rs916069779 Chr.16: 89279509-89279509 on Build GRCh38
rs967759428 Chr.16: 89279444-89279444 on Build GRCh38
rs896510600 Chr.16: 89279483-89279483 on Build GRCh38

View Full Product Details