Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34247342-34247615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf1
Entrez Gene ID
221491
Gene Name
chromosome 6 open reading frame 1
Gene Aliases
LBH
UniGene
Hs.381300
Gene Chromosome Location
Chr.6: 34246380-34249108 on Build GRCh38
Transcripts
NM_001008703.2 NM_001008704.2 NM_001287396.1 NM_001287397.1 NM_001287398.1 NM_178508.4

SNP Detail

SNP ID Chromosome Location
rs867689843 Chr.6: 34247371-34247371 on Build GRCh38
rs773792916 Chr.6: 34247530-34247530 on Build GRCh38
rs986988590 Chr.6: 34247400-34247400 on Build GRCh38
rs769392164 Chr.6: 34247484-34247484 on Build GRCh38
rs781465578 Chr.6: 34247482-34247482 on Build GRCh38
rs1007158885 Chr.6: 34247475-34247475 on Build GRCh38
rs748935629 Chr.6: 34247489-34247489 on Build GRCh38
rs1042908989 Chr.6: 34247498-34247498 on Build GRCh38
rs756622933 Chr.6: 34247422-34247438 on Build GRCh38
rs774315505 Chr.6: 34247418-34247418 on Build GRCh38
rs757767308 Chr.6: 34247477-34247477 on Build GRCh38
rs1026203750 Chr.6: 34247540-34247540 on Build GRCh38
rs928482 Chr.6: 34247444-34247444 on Build GRCh38
rs199814662 Chr.6: 34247524-34247524 on Build GRCh38
rs939225749 Chr.6: 34247373-34247373 on Build GRCh38
rs965619007 Chr.6: 34247515-34247516 on Build GRCh38
rs997181955 Chr.6: 34247595-34247595 on Build GRCh38
rs142296874 Chr.6: 34247483-34247483 on Build GRCh38
rs951512711 Chr.6: 34247561-34247561 on Build GRCh38
rs768192780 Chr.6: 34247516-34247516 on Build GRCh38
rs149332404 Chr.6: 34247407-34247407 on Build GRCh38
rs771403342 Chr.6: 34247565-34247565 on Build GRCh38
rs541128045 Chr.6: 34247442-34247442 on Build GRCh38
rs902595490 Chr.6: 34247521-34247521 on Build GRCh38
rs758471300 Chr.6: 34247464-34247464 on Build GRCh38
rs1055345011 Chr.6: 34247461-34247461 on Build GRCh38
rs752089589 Chr.6: 34247470-34247470 on Build GRCh38
rs1150780 Chr.6: 34247451-34247451 on Build GRCh38
rs759774254 Chr.6: 34247443-34247443 on Build GRCh38
rs138631327 Chr.6: 34247390-34247390 on Build GRCh38
rs984570345 Chr.6: 34247594-34247594 on Build GRCh38
rs761707301 Chr.6: 34247426-34247426 on Build GRCh38
rs1054294860 Chr.6: 34247548-34247548 on Build GRCh38
rs537731519 Chr.6: 34247549-34247549 on Build GRCh38
rs558956626 Chr.6: 34247429-34247429 on Build GRCh38
rs370968291 Chr.6: 34247468-34247468 on Build GRCh38
rs35089390 Chr.6: 34247392-34247393 on Build GRCh38
rs374464893 Chr.6: 34247487-34247487 on Build GRCh38
rs769739811 Chr.6: 34247406-34247406 on Build GRCh38

View Full Product Details