Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 634601-634830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL26
Entrez Gene ID
84326
Gene Name
methyltransferase like 26
Gene Aliases
C16orf13, JFP2
UniGene
Hs.239500
Gene Chromosome Location
Chr.16: 634427-636366 on Build GRCh38
Transcripts
NM_001040160.2 NM_001040161.2 NM_001040162.2 NM_001040165.2 NM_001288710.1 NM_032366.4 NR_109976.1 NR_109977.1 NR_109978.1 NR_109979.1 XM_011522713.2 XM_011522714.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5055024 c.236A>G p.E79G Chr.16: 634759-634759 on Build GRCh38
5055023 c.527A>G p.E176G Chr.16: 634759-634759 on Build GRCh38
3511235 c.540A>C p.E180D Chr.16: 634604-634604 on Build GRCh38
4927959 c.542C>T p.A181V Chr.16: 634744-634744 on Build GRCh38
1708601 c.442G>A p.G148R Chr.16: 634776-634776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758337261 Chr.16: 634752-634752 on Build GRCh38
rs142689363 Chr.16: 634632-634632 on Build GRCh38
rs371245617 Chr.16: 634666-634666 on Build GRCh38
rs200345361 Chr.16: 634807-634807 on Build GRCh38
rs775017055 Chr.16: 634694-634694 on Build GRCh38
rs147359915 Chr.16: 634777-634777 on Build GRCh38
rs894329285 Chr.16: 634734-634734 on Build GRCh38
rs769971293 Chr.16: 634670-634670 on Build GRCh38
rs768035562 Chr.16: 634700-634700 on Build GRCh38
rs761977803 Chr.16: 634788-634788 on Build GRCh38
rs200976892 Chr.16: 634682-634682 on Build GRCh38
rs377633763 Chr.16: 634704-634704 on Build GRCh38
rs750818105 Chr.16: 634702-634702 on Build GRCh38
rs762456139 Chr.16: 634698-634698 on Build GRCh38
rs752173704 Chr.16: 634745-634745 on Build GRCh38
rs922605548 Chr.16: 634641-634641 on Build GRCh38
rs764867078 Chr.16: 634805-634805 on Build GRCh38
rs752560502 Chr.16: 634647-634647 on Build GRCh38
rs772511537 Chr.16: 634791-634791 on Build GRCh38
rs755949121 Chr.16: 634657-634658 on Build GRCh38
rs778444371 Chr.16: 634743-634743 on Build GRCh38
rs942950401 Chr.16: 634624-634624 on Build GRCh38
rs748853423 Chr.16: 634637-634637 on Build GRCh38
rs778853118 Chr.16: 634659-634659 on Build GRCh38
rs148428677 Chr.16: 634794-634794 on Build GRCh38
rs941175257 Chr.16: 634710-634710 on Build GRCh38
rs370828018 Chr.16: 634718-634718 on Build GRCh38
rs189161844 Chr.16: 634669-634669 on Build GRCh38
rs754427197 Chr.16: 634644-634644 on Build GRCh38
rs749468618 Chr.16: 634778-634778 on Build GRCh38
rs777755534 Chr.16: 634754-634754 on Build GRCh38
rs181861375 Chr.16: 634811-634811 on Build GRCh38
rs765960949 Chr.16: 634705-634705 on Build GRCh38
rs753449471 Chr.16: 634712-634712 on Build GRCh38
rs759228854 Chr.16: 634803-634803 on Build GRCh38
rs774610311 Chr.16: 634782-634782 on Build GRCh38
rs373878833 Chr.16: 634769-634769 on Build GRCh38
rs760952446 Chr.16: 634801-634801 on Build GRCh38
rs764225691 Chr.16: 634687-634687 on Build GRCh38
rs779751719 Chr.16: 634635-634635 on Build GRCh38
rs377689531 Chr.16: 634663-634663 on Build GRCh38
rs370300432 Chr.16: 634667-634667 on Build GRCh38
rs36022017 Chr.16: 634731-634732 on Build GRCh38
rs746797280 Chr.16: 634760-634760 on Build GRCh38
rs373936560 Chr.16: 634627-634627 on Build GRCh38
rs1024645244 Chr.16: 634790-634790 on Build GRCh38
rs754180031 Chr.16: 634802-634802 on Build GRCh38
rs775727636 Chr.16: 634679-634679 on Build GRCh38

View Full Product Details