Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 18657566-18657839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
UniGene
Hs.741483
Gene Chromosome Location
Chr.9: 17906574-18910950 on Build GRCh38
Transcripts
NM_001040272.5 NM_052866.4 XM_011518063.2 XM_011518064.2 XM_017015310.1 XM_017015311.1 XM_017015312.1 XM_017015313.1 XM_017015314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6334195 c.848G>T p.G283V Chr.9: 18657652-18657652 on Build GRCh38
4431787 c.909G>A p.E303E Chr.9: 18657713-18657713 on Build GRCh38
6473067 c.919T>C p.F307L Chr.9: 18657723-18657723 on Build GRCh38
6594401 c.918C>A p.F306L Chr.9: 18657722-18657722 on Build GRCh38
7282025 c.839G>A p.R280H Chr.9: 18657643-18657643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750236831 Chr.9: 18657643-18657643 on Build GRCh38
rs374986484 Chr.9: 18657734-18657734 on Build GRCh38
rs777979788 Chr.9: 18657777-18657777 on Build GRCh38
rs369647812 Chr.9: 18657791-18657791 on Build GRCh38
rs557986753 Chr.9: 18657627-18657627 on Build GRCh38
rs964034400 Chr.9: 18657796-18657796 on Build GRCh38
rs765849326 Chr.9: 18657598-18657598 on Build GRCh38
rs765103865 Chr.9: 18657632-18657632 on Build GRCh38
rs370950060 Chr.9: 18657596-18657596 on Build GRCh38
rs375693358 Chr.9: 18657603-18657603 on Build GRCh38
rs890950152 Chr.9: 18657716-18657716 on Build GRCh38
rs267602193 Chr.9: 18657658-18657658 on Build GRCh38
rs772334180 Chr.9: 18657772-18657772 on Build GRCh38
rs756550894 Chr.9: 18657730-18657730 on Build GRCh38
rs770791137 Chr.9: 18657666-18657666 on Build GRCh38
rs768827111 Chr.9: 18657589-18657589 on Build GRCh38
rs555934579 Chr.9: 18657606-18657606 on Build GRCh38
rs760606499 Chr.9: 18657660-18657660 on Build GRCh38
rs775124323 Chr.9: 18657793-18657793 on Build GRCh38
rs764741306 Chr.9: 18657609-18657609 on Build GRCh38
rs767980152 Chr.9: 18657725-18657725 on Build GRCh38
rs372646636 Chr.9: 18657705-18657705 on Build GRCh38
rs773037108 Chr.9: 18657659-18657659 on Build GRCh38
rs192634806 Chr.9: 18657800-18657800 on Build GRCh38
rs776395871 Chr.9: 18657784-18657784 on Build GRCh38
rs754970661 Chr.9: 18657757-18657757 on Build GRCh38
rs776647405 Chr.9: 18657676-18657676 on Build GRCh38
rs758182595 Chr.9: 18657728-18657728 on Build GRCh38
rs550978123 Chr.9: 18657759-18657759 on Build GRCh38
rs752262465 Chr.9: 18657703-18657703 on Build GRCh38
rs368670565 Chr.9: 18657649-18657649 on Build GRCh38
rs769166422 Chr.9: 18657792-18657792 on Build GRCh38
rs764835876 Chr.9: 18657686-18657686 on Build GRCh38
rs372263270 Chr.9: 18657591-18657591 on Build GRCh38
rs561847044 Chr.9: 18657715-18657715 on Build GRCh38
rs201832138 Chr.9: 18657743-18657743 on Build GRCh38
rs201308391 Chr.9: 18657642-18657642 on Build GRCh38
rs376640618 Chr.9: 18657592-18657592 on Build GRCh38
rs544264092 Chr.9: 18657760-18657760 on Build GRCh38
rs768793788 Chr.9: 18657650-18657650 on Build GRCh38
rs187650110 Chr.9: 18657630-18657630 on Build GRCh38
rs374914646 Chr.9: 18657778-18657778 on Build GRCh38
rs748344122 Chr.9: 18657769-18657769 on Build GRCh38
rs748431522 Chr.9: 18657657-18657657 on Build GRCh38
rs1038312943 Chr.9: 18657639-18657639 on Build GRCh38
rs139269453 Chr.9: 18657656-18657656 on Build GRCh38
rs936804520 Chr.9: 18657781-18657781 on Build GRCh38
rs371712616 Chr.9: 18657756-18657756 on Build GRCh38
rs770785740 Chr.9: 18657780-18657780 on Build GRCh38
rs751197103 Chr.9: 18657804-18657804 on Build GRCh38
rs767320607 Chr.9: 18657593-18657593 on Build GRCh38
rs746356399 Chr.9: 18657634-18657634 on Build GRCh38
rs546088260 Chr.9: 18657597-18657597 on Build GRCh38
rs776558387 Chr.9: 18657585-18657585 on Build GRCh38

View Full Product Details